Skip to Content
Merck
All Photos(1)

Documents

438626

Sigma-Aldrich

4-Hydroxy-3-nitrocoumarin

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5NO5
CAS Number:
Molecular Weight:
207.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

172 °C (dec.) (lit.)

SMILES string

OC1=C(C(=O)Oc2ccccc12)[N+]([O-])=O

InChI

1S/C9H5NO5/c11-8-5-3-1-2-4-6(5)15-9(12)7(8)10(13)14/h1-4,11H

InChI key

NZQAQAUWFHMVEM-UHFFFAOYSA-N

General description

4-Hydroxy-3-nitrocoumarin is a coumarin derivative and its cytotoxic action against cultured human tumor and normal cells has been investigated. It can be prepared by the nitration of 4-hydroxycoumarin in glacial acetic acid by using 72% HNO3.

Application

4-Hydroxy-3-nitrocoumarin may be used as starting reagent for the synthesis of following compounds:
  • 4-chloro-3-nitrocoumarin
  • 2-unsubstituted 3-nitrochromone
  • 4-amino-3-nitrocoumarins

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masami Kawase et al.
In vivo (Athens, Greece), 19(4), 705-711 (2005-07-08)
A preliminary exploration of coumarin derivatives as novel multidrug resistance (MDR) modulators was carried out to determine the basic features of the structure responsible for the MDR reversal activity. Among 44 coumarins, 14 compounds moderately induced the reversal of MDR
Biljana R Dekić et al.
Molecules (Basel, Switzerland), 15(4), 2246-2256 (2010-04-30)
Synthesis, spectral analysis and bioactivity of new coumarin derivatives are described in this paper. Eight new coumarin derivatives were synthesized in moderate to good yields by condensation of 4-chloro-3-nitrocoumarin and the corresponding heteroarylamine. The synthesized compounds were tested for their
Synthesis of heteroannulated 3-nitro-and 3-aminopyridines by cyclocondensation of electron-rich aminoheterocycles with 3-nitrochromone.
Iaroshenko VO, et al.
Tetrahedron, 68(11), 2532-2543 (2012)
Investigations of pyrans and related compounds.
Savel'ev VL, et al.
Chemistry of Heterocyclic Compounds, 9(7), 816-820 (1973)
Vidoslav Dekić et al.
Magnetic resonance in chemistry : MRC, 48(11), 896-902 (2010-09-08)
Herein, we describe the synthesis and complete assignment of the (1)H and (13)C NMR chemical shifts of a series of antimicrobial 4-arylamino-3-nitrocoumarin derivatives based on a combination of (1)H and (13)C NMR, (1)H-(1)H-COSY, NOESY, HSQC and HMBC experiments. Conformational effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service