Skip to Content
Merck
All Photos(1)

Documents

414131

Sigma-Aldrich

1-Bromopinacolone

≥97%

Synonym(s):

1-Bromo-3,3-dimethyl-2-butanone, Bromomethyl tert-butyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCOCH2Br
CAS Number:
Molecular Weight:
179.05
Beilstein:
506485
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

form

liquid

refractive index

n20/D 1.466 (lit.)

bp

188-194 °C (lit.)

mp

−10 °C (lit.)

density

1.331 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CC(C)(C)C(=O)CBr

InChI

1S/C6H11BrO/c1-6(2,3)5(8)4-7/h4H2,1-3H3

InChI key

SAIRZMWXVJEBMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromopinacolone is an α-bromoketone-aldehyde. Coupling of 1-bromopinacolone with various aldehyde electrophiles catalyzed by SmI2 has been reported. It acts as reversible competitive inhibitor for acetylcholinesterase, during hydrolysis of acetylcholine.

Application

1-Bromopinacolone is the suitable reagent for the synthesis of a photolabile azido derivative of the kaurene oxidase inhibitor 1-(4- chlorophenyl)4,4-dimethyl-2-(1,2,4-triazol-1-yl) pentan-3-ol (paclobutrazol). It may be used as reagent in the synthesis of 2-t-butyl-6-benzoylimidazo[1,2-b]pyridazine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian A Sparling et al.
Organic letters, 10(6), 1291-1294 (2008-02-28)
Highly substituted, very hindered enones were synthesized using a two-step procedure that utilizes a diiodosamarium-promoted Reformatsky-type coupling and dehydration using Martin sulfurane. Both alpha-chloro- and alpha-bromoketones were coupled with a variety of carbonyl nucleophiles to form the intermediate beta-hydroxyketones, occurring
D L Hallahan et al.
Plant physiology, 88(4), 1425-1429 (1988-12-01)
A photolabile azido derivative of the kaurene oxidase inhibitor 1-(4-chlorophenyl)-4,4-dimethyl-2-(1,2,4-triazol-l-yl) pentan-3-ol (paclobutrazol) has been synthesized for use as a photoaffinity labeling agent. The compound was tested as an inhibitor of the oxidation of ent-kaurene catalyzed by cell-free preparations from endosperm
Methyl imidazo [1,2-b] pyridazine-2-carbamates and related compounds as potential antifilarial agents
Mourad AE, et al.
Journal of Heterocyclic Chemistry, 29, 1583-1583 (1992)
S G Cohen et al.
Biochimica et biophysica acta, 997(3), 167-175 (1989-08-31)
1-Bromo-2-[14C]pinacolone, (CH3)3C14COCH2Br [( 14C]BrPin), was prepared from [1-14C]acetyl chloride and tert-butylmagnesium chloride with cuprous chloride catalyst, followed by bromination. It was examined as an active-site directed label for acetylcholinesterase (acetylcholine acetylhydrolase, EC 3.1.1.7) (AcChE). AcChE, isolated from Torpedo nobiliana, has
E Salih et al.
The Journal of biological chemistry, 268(1), 245-251 (1993-01-05)
Acetylcholinesterase (AcChE, EC 3.1.1.7) was isolated from the electric organ of T. nobiliana and treated with the active-site-directed alkylating agent 1-bromo-2-[14C]pinacolone ([14C]BrPin), or with BrPin, which acts initially as a competitive inhibitor, Ki = 0.18 mM, and then inactivates the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service