Skip to Content
Merck
All Photos(2)

Documents

412449

Sigma-Aldrich

PAMAM dendrimer, ethylenediamine core, generation 4.0 solution

10 wt. % in methanol

Synonym(s):

Starburst Generation 4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=4);dendri PAMAM(NH2)64
CAS Number:
Molecular Weight:
14214.17
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

amino surface groups

form

solution

mol wt

generation 4.0

feature

core type ethylenediamine core (2-carbon core)

concentration

10 wt. % in methanol

no. Surface Groups

64

refractive index

n20/D 1.348

bp

64.7 °C

density

0.813 g/mL at 25 °C

storage temp.

2-8°C

General description

PAMAM dendrimer, ethylenediamine core, generation 4.0 solution is a poly(amidoamine) that is categorized as a class of dendritic polymers which can be prepared by a two-step interactive reaction that forms generations around the central ethylenediamine core. It consists of nitrogen and amide as functional groups that are structured in regular “branched upon branched” pattern. It is majorly used as a high capacity chelating agent.

Application

PAMAM dendrimer, ethylenediamine core, generation 4.0 solution is used as a compacting agent to bring in the conformational changes in the DNA molecule. It can be attached on the cellulose substrate that acts as a biofunctional surface for potential applications in immonoassays. It is used in MRI contrast agents and in the condensation formation of silica nanospheres.

Physical form

Contains 64 surface primary amino groups

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felipe Vidal et al.
Molecular pharmaceutics, 13(10), 3395-3403 (2016-08-25)
Polyamidoamine (PAMAM) dendrimers are hyperbranched macromolecules which have been described as one of the most promising drug nanocarrier systems. A key process to understand is their cellular internalization mechanism because of its direct influence on their intracellular distribution, association with
DNA compaction induced by a cationic polymer or surfactant impact gene expression and DNA degradation
Ainalem M, et al.
PLoS ONE, 9(3), e92692-e92692 (2014)
Rolando Alberto Rodríguez-Fonseca et al.
Colloids and surfaces. B, Biointerfaces, 177, 77-93 (2019-02-04)
Peptide epitopes have been widely used to develop synthetic vaccines and immunotherapies. However, peptide epitopes may exhibit poor absorption or immunogenicity due to their low molecular weights. Conversely, fourth-generation polyamidoamine (G4-PAMAM) dendrimers are nonimmunogenic and relatively nontoxic synthetic nanoparticles that
Marta Walczynska et al.
Toxicology mechanisms and methods, 28(6), 432-439 (2018-04-03)
Nematode Caenorhabditis elegans (C. elegans) was used to investigate the impact of silver nanoparticles (SNP), multiwalled carbon nanotubes (MWCNT), and polyamidoamine dendrimers (PAMAM) used in concentration of 1010 particle/mL. Population-based observations and gene expression analysis were employed in this study.
Dendritic chelating agents. 1. Cu (II) binding to ethylene diamine core poly (amidoamine) dendrimers in aqueous solutions
Diallo MS, et al.
Langmuir, 20(7), 2640-2651 (2004)

Articles

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service