Skip to Content
Merck
All Photos(1)

Key Documents

40477

Sigma-Aldrich

1-Methylimidazolium chloride

95%

Synonym(s):

1-Methylimidazole hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2 · HCl
CAS Number:
Molecular Weight:
118.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥94.5% (HPLC)
95%

impurities

≤1% water

SMILES string

Cl[H].Cn1ccnc1

InChI

1S/C4H6N2.ClH/c1-6-3-2-5-4-6;/h2-4H,1H3;1H

InChI key

STCBHSHARMAIOM-UHFFFAOYSA-N

Application

1-Methylimidazolium chloride behaves as a solvent and catalyst during the formation of 5-hydroxymethylfurfural from fructose and sucrose via acid-catalyzed dehydration.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Najeh Maaloul et al.
Environmental science and pollution research international, 27(19), 23447-23463 (2019-01-04)
In this study, novel cellulose-bead-based biosorbents (CBBAS) were successfully synthesized from almond shell using a simple three-step process: (i) dissolution of bleached almond shell in ionic liquid (1-butyl-3-methylimidazolium chloride), (ii) coagulation of cellulose-ionic liquid solution in water and (iii) freeze-drying.
Pankaj Attri et al.
PloS one, 8(7), e68970-e68970 (2013-07-23)
In this paper, we have examined the conductivity and interaction studies of ammonium and imidazolium based ionic liquids (ILs) with the newly synthesised low bandgap polymer (Poly(2-heptadecyl-4-vinylthieno[3,4-d]thiazole) (PHVTT)). Use of low bandgap polymers is the most suitable way to harvest
Zhuang Wang et al.
Chemosphere, 185, 681-689 (2017-07-21)
Graphene and ionic liquids (ILs) released into the environment will interact with each other. So far however, the risks associated with the concurrent exposure of biota to graphene and ILs in the environment have received little attention. The research reported
Ariyanti Sarwono et al.
Ultrasonics sonochemistry, 37, 310-319 (2017-04-22)
5-Hydroxymethylfurfural (HMF) has been identified as a promising biomass-derived platform chemical. In this study, one pot production of HMF was studied in ionic liquid (IL) under probe sonication technique. Compared with the conventional heating technique, the use of probe ultrasonic
Dehydration of fructose and sucrose into 5-hydroxymethylfurfural in the presence of 1-H-3-methyl imidazolium chloride acting both as solvent and catalyst.
Moreau C, et al.
J. Mol. Catal. A: Chem., 253(1), 165-169 (2006)

Articles

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service