Skip to Content
Merck
All Photos(1)

Documents

396540

Sigma-Aldrich

1,2-Dichloroethane-d4

99 atom % D

Synonym(s):

Ethylene-d4 dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCD2CD2Cl
CAS Number:
Molecular Weight:
102.98
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99 atom % D

Assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.443 (lit.)

bp

83 °C (lit.)

mp

−35 °C (lit.)

density

1.307 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

mass shift

M+4

SMILES string

[2H]C([2H])(Cl)C([2H])([2H])Cl

InChI

1S/C2H4Cl2/c3-1-2-4/h1-2H2/i1D2,2D2

InChI key

WSLDOOZREJYCGB-LNLMKGTHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Dichloroethane-d4 is 1,2-dichloroethane with all the hydrogen atoms replaced by deuterium. Its synthesis and structure elucidation by neutron diffraction measurements has been reported.

Application

1,2-Dichloroethane-d4 may be used:
  • As a NMR solvent.
  • As a surrogate standard during the analysis of volatile organic compounds (VOC) in water by gas chromatography-mass spectrometry (GC-MS).
  • As a chain termination reagent in the synthesis of poly(ethylene-d4 oxide).

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Asp. Tox. 1 - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.8 °F - closed cup - Based on data from similar materials

Flash Point(C)

16.0 °C - closed cup - Based on data from similar materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An automated GC/MS system for the analysis of volatile and semi-volatile organic compounds in water.
Vickers AK and Wright LM.
Journal of Analytical Methods in Chemistry, 15(4), 133-139 (1993)
Volatile organic compounds in water near petrochemical factories in Taiwan.
Kuo HW, et al.
Chemosphere, 33(5), 913-920 (1996)
Solvent Effect on Chiral Aggregate Formation of Acylamino Acids.
Iwahashi M, et al.
Journal of Oleo Science, 51(11), 705-713 (2002)
Synthesis of poly (ethylene-d4oxide) with unimodal and narrow molecular weight distribution.
Eisenbach CD, et al.
Macromolecular Chemistry and Physics, 184(11), 2313-2323 (1983)
The structure of liquid 1, 2 dichloroethane by neutron diffraction. I. Molecular structure, full pair correlation function, and temperature effects.
Bermejo FJ, et al.
J. Chem. Phys. , 87(12), 7171-7178 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service