Skip to Content
Merck
All Photos(2)

Key Documents

39410

Sigma-Aldrich

4-(Dimethylamino)pyridine, polymer-bound

extent of labeling: ~3.0 mmol/g "DMAP" loading, matrix crosslinked with 2% DVB

Synonym(s):

4-(Dimethylamino)pyridine on Polystyrene, DMAP on PS, polymer bound DMAP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8
CAS Number:
Molecular Weight:
104.15
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

~3.0 mmol/g "DMAP" loading

matrix

crosslinked with 2% DVB

anion traces

chloride (Cl-): ≤5000 mg/kg

Looking for similar products? Visit Product Comparison Guide

General description

Polymer-bound 4-(dimethylamino)pyridine is used for improved derivatization of chiral and achiral aliphatic amines, amino alcohols and amino acids.

Application

Polymer-bound 4-(dimethylamino)pyridine is used as a supported base in the Michael addition and nitrous acid elimination process step involved in the synthesis of thiophene-2-carboxylates using β-nitroacrylates as starting materials, under a general and efficient continuous flow-based protocol. It also serves as a supported catalyst in the preparation of ω-nitro esters starting from cyclic 2-nitro ketones under solid heterogeneous catalysis.(5)

Other Notes

Polymer supported, recyclable acylation catalystcitationcitationcitation
Polymer supported, recyclable acylation catalyst

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

$\beta$-Nitroacrylates as Starting Materials of Thiophene-2-Carboxylates Under Continuous Flow Conditions
Chiurchiu E, et al.
Advanced Synthesis & Catalysis, 361(9), 2042-2047 (2019)
A new, low impact and efficient synthesis of $\omega$-nitro esters under solid heterogeneous catalysis
Chiurchiu E, et al.
Green Chemistry, 19(20), 4956-4960 (2017)
$\beta$-Nitroacrylates as Starting Materials of Thiophene-2-Carboxylates Under Continuous Flow Conditions
Chiurchiu E, et al.
advanced synthesis and catalysis, 361(9), 2042-2047 (2019)
Szu-Yuan Chou et al.
Scientific reports, 10(1), 18519-18519 (2020-10-30)
Cell development and behavior are driven by internal genetic programming, but the external microenvironment is increasingly recognized as a significant factor in cell differentiation, migration, and in the case of cancer, metastatic progression. Yet it remains unclear how the microenvironment
E.F. Scriven
Chemical Society Reviews, 12, 129-129 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service