392855
4,5-Dimethoxy-2-nitrobenzyl bromide
97%
Synonym(s):
6-Nitroveratryl bromide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
O2NC6H2(OCH3)2CH2Br
CAS Number:
Molecular Weight:
276.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
131-133 °C (lit.)
functional group
bromo
nitro
SMILES string
COc1cc(CBr)c(cc1OC)[N+]([O-])=O
InChI
1S/C9H10BrNO4/c1-14-8-3-6(5-10)7(11(12)13)4-9(8)15-2/h3-4H,5H2,1-2H3
InChI key
UEKFEYNZISYRRH-UHFFFAOYSA-N
Related Categories
General description
4,5-Dimethoxy-2-nitrobenzyl bromide (DMNBB, 1-(Bromomethyl)-4,5-dimethoxy-2-nitrobenzene) is a 4,5-dimethoxy-2-nitrobenzyl derivative. 4,5-Dimethoxy-2-nitrobenzyl (DMNB) group of DMNBB is used as a photolabile protecting group in caging technology to develop pro-drugs. Synthesis of 1-(bromomethyl)-4,5-dimethoxy-2-nitrobenzene by using 6-nitroveratraldehyde as starting reagent has been reported.
Application
4,5-Dimethoxy-2-nitrobenzyl bromide (DMNB bromide, 6-nitroveratryl bromide) is suitable reagent used in the synthesis of N-(4,5-dimethoxy-2-nitrobenzyl)vanillylamine which forms caged vanilloid. It may be used in the synthesis of the following:
- 4-(4′,5′-dimethoxy-2-nitrobenzyloxy)benzaldehyde, a DMNB-caged aldehyde
- N-[4-[(4,5-dimethoxy-2-nitrobenzyl)oxy]-3-methoxybenzyl]acetamide
- caged derivative of pyridostatin ([C]-PDS)
- photosensitive polyimide (PI-DMNB)
- caged β-ecdysone
- 4-tert-butyldimethylsilyloxy-1-(2-deoxy-3,5-di-O-toluoyl-β-D-ribofuranosyl)-2-(6-nitroveratrylthio)-1H-benzimidazole, an intermediate in synthesis of phosphoramidite bearing 4-hydroxy-2-mercaptobenzimidazole (SBNV) nucleobase
- alkylation of dihydrofluorescein
- 4-(4′,5′-Dimethoxy-2′-nitrobenzyloxy)benzaldehyde
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
G Marriott et al.
Biochemistry, 35(10), 3170-3174 (1996-03-12)
An understanding of the molecular mechanism of muscle contraction will require a complete description of the kinetics of the myosin motor in vitro and in vivo. To this end chemical relaxation studies employing light-directed generation of ATP from caged ATP
Surface functionalization of PEEK films using photochemical routes.
Henneuse-Boxus C, et al.
Eur. Polymer J., 37(1), 9-18 (2001)
Nicolaos Avlonitis et al.
Molecular bioSystems, 5(5), 450-457 (2009-04-22)
The study of mitochondria and mitochondrial Ca2+ signalling in localised regions is hampered by the lack of tools that can uncouple the mitochondrial membrane potential (DeltaPsi(m)) in a spatially predefined manner. Although there are a number of existing mitochondrial uncouplers
Luke D Lavis et al.
ACS chemical biology, 9(4), 855-866 (2014-03-04)
Small-molecule fluorophores manifest the ability of chemistry to solve problems in biology. As we noted in a previous review (Lavis, L. D.; Raines, R. T. ACS Chem. Biol. 2008, 3, 142-155), the extant collection of fluorescent probes is built on
Synthesis and Characterizations of Positive-Working Photosensitive Polyimides Having 4, 5-Dimethoxy-o-Nitrobenzyl Side Group.
Ryu S, et al.
Bull. Korean Chem. Soc., 29(9), 1689-1689 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service