Skip to Content
Merck
All Photos(1)

Key Documents

392510

Sigma-Aldrich

1-Iodo-3,5-dimethylbenzene

99%

Synonym(s):

5-Iodo-m-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3I
CAS Number:
Molecular Weight:
232.06
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.594 (lit.)

bp

92-94 °C/3 mmHg (lit.)

density

1.608 g/mL at 25 °C (lit.)

functional group

iodo

SMILES string

Cc1cc(C)cc(I)c1

InChI

1S/C8H9I/c1-6-3-7(2)5-8(9)4-6/h3-5H,1-2H3

InChI key

ZLMKEENUYIUKKC-UHFFFAOYSA-N

General description

1-Iodo-3,5-dimethylbenzene (5-Iodo-m-xylene) is an aryl halide. It can be obtained from 5-bromo-m-xylene, via copper-catalyzed halogen exchange reaction, in the presence of NaI or KI in n-BuOH or DMF (solvents). It undergoes reaction with phenol in the presence of CuFe2O4 nano powder as a recyclable catalyst to afford 1,3-dimethyl-5-phenoxybenzene.

Application

1-Iodo-3,5-dimethylbenzene (5-iodo-m-xylene) is suitable for use in the synthesis of N-(3,5-xylyl)-N-ethylaniline, an arylamine.
It may be used in the following studies:
  • α-Arylation of ketones.
  • Copper-catalyzed N-arylation of imidazoles.
  • Cyanation of 5-iodo-m-xylene to form 3,5-dimethylbenzonitrile.
  • Synthesis of 1,3-Dimethyl-5-phenoxybenzene by nano-CuFe2O4 catalyzed C-O cross-coupling with phenol.
  • CuBr-catalyzed amination of 1-iodo-3,5-dimethylbenzene to form N-Allyl-3,5-dimethylbenzenamine.
  • Copper-catalyzed C-S bond-formation between 5-iodo-m-xylene and thiophenol.
  • As a starting material in the synthesis of biphenyl-3,3′,5,5′-tetracarboxylic acid.
  • Radical bromination of 5-iodo-m-xylene by N-bromosuccinimide to form 1,3-bis(bromomethyl)-5-iodobenzene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M H Ali et al.
The Journal of organic chemistry, 66(8), 2560-2565 (2001-04-17)
Aryl iodides are coupled with amines to give the corresponding arylamines in high yield in the presence of palladium, a suitable ligand, and NaOt-Bu. Functionalized aryl iodides give good yields of the corresponding arylamines when Cs(2)CO(3) is substituted as the
Biphenyl-3, 3', 5, 5'-tetracarboxylic acid.
Coles SJ, et al.
Acta Crystallographica Section E, Structure Reports Online, 58(6), 626-628 (2002)
On the synthesis of heterocyclic dendrons.
Diez-Barra E, et al.
ARKIVOC (Gainesville, FL, United States), 2002(5), 17-25 (2002)
Fuk Yee Kwong et al.
Organic letters, 4(20), 3517-3520 (2002-09-27)
An efficient copper-catalyzed carbon-sulfur bond formation reaction was developed. This method is particularly noteworthy given its experimental simplicity, high generality, and exceptional level of functional group toleration and the low cost of the catalyst system. [reaction: see text]
Anouk S Lubbe et al.
The Journal of organic chemistry, 76(21), 8599-8610 (2011-09-21)
A study is presented on the control of rotary motion of an appending rotor unit in a light-driven molecular motor. Two new light driven molecular motors were synthesized that contain aryl groups connected to the stereogenic centers. The aryl groups

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service