Skip to Content
Merck
All Photos(1)

Key Documents

392103

Sigma-Aldrich

Isobornyl acrylate

technical grade, contains 200 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

Acrylic acid isobornyl ester, IBA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H20O2
CAS Number:
Molecular Weight:
208.30
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

form

liquid

contains

200 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.476 (lit.)

bp

119-121 °C/15 mmHg (lit.)

density

0.986 g/mL at 25 °C (lit.)

SMILES string

[H][C@]12CC[C@@](C)([C@@H](C1)OC(=O)C=C)C2(C)C

InChI

1S/C13H20O2/c1-5-11(14)15-10-8-9-6-7-13(10,4)12(9,2)3/h5,9-10H,1,6-8H2,2-4H3/t9-,10-,13+/m0/s1

InChI key

PSGCQDPCAWOCSH-OUJBWJOFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isobornyl acrylate (IA) is an acrylate-based monomer used in the production of various polymers, coatings, and adhesives. It has excellent adhesion properties, good chemical resistance, and low shrinkage, making it a popular choice for use in UV-curable coatings on various substrates such as plastics, metals, and wood. Additionally, it is also used as a crosslinking agent in radiation-curable coatings and as an additive for pressure-sensitive adhesives.

Application

Isobornyl acrylate can be used as a co-monomer with other acrylic monomers in the preparation of pressure-sensitive adhesives (PSAs) for optical applications. It is also used in the development of high-performance and stable polymer gate dielectrics for organic thin-film transistors (OTFTs). Furthermore, it can also used in the preparation of UV-curable waterborne polyurethane (WPU) by copolymerization.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hee-Woong Park et al.
Polymers, 12(10) (2020-10-23)
To improve the heat resistance of acrylic-based pressure-sensitive adhesive (PSA), silicone-block-containing acrylic PSAs (SPSAs) were synthesized using a polydimethylsiloxane (PDMS)-based macro-azo-initiator (MAI). To evaluate the heat resistance of the PSA films, the probe tack and 90° peel strength were measured
Yih-Lin Cheng et al.
Polymers, 12(3) (2020-03-18)
Material jetting (MJ)-type 3D printers have been considered as one of the most versatile types of 3D printers, enabling full-color printing and multi-material printing. However, to the best of our knowledge, there are few academic studies on the development of
A Mesgarnejad et al.
Physical review. E, 102(1-1), 013004-013004 (2020-08-17)
While cracks in isotropic homogeneous materials propagate straight, perpendicularly to the tensile axis, cracks in natural and synthetic composites deflect from a straight path, often increasing the toughness of the material. Here we combine experiments and simulations to identify materials
Comparison of thermomechanical properties of statistical, gradient and block copolymers of isobornyl acrylate and n-butyl acrylate with various acrylate homopolymers
Jakubowski W, et al.
Polymer, 49(6), 1567-1578 (2008)
Huawei Jiang et al.
Scientific reports, 7, 41208-41208 (2017-01-26)
This paper reports on a miniaturized microbial fuel cell with a microfluidic flow-through configuration: a porous anolyte chamber is formed by filling a microfluidic chamber with three-dimensional graphene foam as anode, allowing nutritional medium to flow through the chamber to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service