Skip to Content
Merck
All Photos(1)

Documents

390593

Sigma-Aldrich

1-(1-Adamantyl)ethylamine hydrochloride

99%

Synonym(s):

α-Methyltricyclo[3.3.1.13,7]-decane-1-methanamine hydrochloride, Rimantadine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21N · HCl
CAS Number:
Molecular Weight:
215.76
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

>300 °C (lit.)

SMILES string

Cl.C[C@H](N)C12C[C@H]3C[C@H](C[C@H](C3)C1)C2

InChI

1S/C12H21N.ClH/c1-8(13)12-5-9-2-10(6-12)4-11(3-9)7-12;/h8-11H,2-7,13H2,1H3;1H/t8?,9-,10+,11-,12-;

InChI key

OZBDFBJXRJWNAV-MZHJCFSPSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G G U Rohde
Der Internist, 52(9), 1047-1052 (2011-08-03)
Influenza infections have important socio-economic consequences. Risk groups identified so far include small children and elderly adults with comorbidities. In recent years in addition to seasonal influenza an outbreak of avian influenza occurred in 2005 and the new H1N1 pandemic
Guang-Quan Wang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 98, 275-281 (2012-09-11)
Amantadine hydrochloride (AMA) and rimantadine hydrochloride (RIM) are non-fluorescent in aqueous solutions. This property makes their determination through direct fluorescent method difficult. The competing reactions and the supramolecular interaction mechanisms between the two drugs and coptisine (COP) as they fight
Rafal M Pielak et al.
Structure (London, England : 1993), 19(11), 1655-1663 (2011-11-15)
The M2 channel of influenza A is a target of the adamantane family antiviral drugs. Two different drug-binding sites have been reported: one inside the pore, and the other is a lipid-facing pocket. A previous study showed that a chimera
E I Burtseva et al.
Voprosy virusologii, 57(1), 20-28 (2012-05-26)
The paper gives the results of monitoring the circulation of influenza viruses in the 2010-2011 season, that covers the second year of circulation of pandemic A(H1N1)v virus strains, and their interaction with seasonal A (H3N2) and B strains. Unlike the
Linh Tran et al.
Molecules (Basel, Switzerland), 16(12), 10227-10255 (2011-12-14)
The M2 channel protein on the influenza A virus membrane has become the main target of the anti-flu drugs amantadine and rimantadine. The structure of the M2 channel proteins of the H3N2 (PDB code 2RLF) and 2009-H1N1 (Genbank accession number

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service