Skip to Content
Merck
All Photos(2)

Key Documents

386529

Sigma-Aldrich

Chlorotrimethylsilane

purified by redistillation, ≥99%

Synonym(s):

TMSCl, TMCS, Trimethylchlorosilane, Trimethylsilyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCl
CAS Number:
Molecular Weight:
108.64
Beilstein:
1209232
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.7 (vs air)

Quality Level

vapor pressure

100 mmHg ( 25 °C)

Assay

≥99%

form

liquid

autoignition temp.

752 °F

purified by

glass distillation
redistillation

expl. lim.

6.4 %

impurities

<0.1% dichlorodimethylsilane

refractive index

n20/D 1.387 (lit.)

bp

57 °C (lit.)

mp

−40 °C (lit.)

density

0.856 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)Cl

InChI

1S/C3H9ClSi/c1-5(2,3)4/h1-3H3

InChI key

IJOOHPMOJXWVHK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorotrimethylsilane is a chloroorganosilane compound mainly used for silylation reactions.

Application

Chlorotrimethylsilane (TMSCl) can be used as:       
  • A reagent to protect alcohol and amine groups via the formation of trimethylsilyl ethers and trimethylsilyl amines.      
  • A catalyst for the preparation of 1,3-diphenyl-2-propenone derivatives (chalcones) as antimicrobial agents.        
  • A trapping agent for the anions generated during acyloin condensation reaction.       
  • A better alternative catalyst to the toxic mercuric chloride for the activation of samarium (Sm) during the cyclopropanation of both allylic and α-allenic alcohols.
  •  A catalyst in the transesterification of triglycerides with alcohols to form fatty acid alkyl esters.        
  • A reagent along with lithium bromide for the conversion of alcohols to the corresponding bromides.       
  • A reagent in Fischer glycosidation.       
  • A source of acid catalyst in the reductive benzylation reaction using benzaldehyde and Et3SiH.
  • A reagent to synthesize sodium trimethylsilanethiolate (Me3SiSNa) by reacting with sodium sulfide, which is an odorless alternative method of synthesizing Me3SiSNa from foul-smelling bis(trimethylsilyl)sulfide and sodium methoxide.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lijie Zhou et al.
Polymers, 11(4) (2019-04-21)
Under the current situation of frequent oil spills, the development of green and recyclable high-efficiency oil-absorbing aerogel materials has attracted wide attention from researchers. In this study, we report a high-strength, three-dimensional hydrophobic cellulose nanofiber (CNF)/polyvinyl alcohol (PVA)/graphene oxide (GO)
Chlorotrimethylsilane as a mild and effective source of acid catalyst in reductive benzylation
Izumi M and Fukase K
Chemistry Letters (Jpn), 34(4), 594-595 (2005)
Potassium dodecatangestocobaltate trihydrate (K5CoW12O40{\textperiodcentered} 3H2O): a mild and efficient catalyst for deprotection of dioxolanes and trimethylsilyl ethers
Habibi MH, et al.
Tetrahedron Letters, 42(38), 6771-6774 (2001)
Guozhong Zhao et al.
Frontiers in microbiology, 11, 579808-579808 (2021-01-26)
Japanese soy sauce has become more acceptable by Chinese consumers due to its umami taste. However, the volatile flavor compounds and taste characters have not been fully clarified. This study aimed to explore the flavor characteristics of three kinds of
Houben-Weyl Methods of Molecular Transformations Vol. 4: Compounds of Groups 15 (As, Sb, Bi) and Silicon Compounds
Sci. Synth., 261-261 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service