Skip to Content
Merck
All Photos(2)

Documents

383473

Sigma-Aldrich

3-Formyl-6-methylchromone

97%

Synonym(s):

6-Methyl-4-oxo-4H-1-benzopyran-3-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8O3
CAS Number:
Molecular Weight:
188.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

172-173 °C (lit.)

SMILES string

Cc1ccc2OC=C(C=O)C(=O)c2c1

InChI

1S/C11H8O3/c1-7-2-3-10-9(4-7)11(13)8(5-12)6-14-10/h2-6H,1H3

InChI key

GBWMIOYSMWCYIZ-UHFFFAOYSA-N

General description

3-Formyl-6-methylchromone is a 3-formyl-substituted chromone. The combustion calorimetric estimation of the enthalpy of combustion of 3-formyl-6-methylchromone has been reported. Its anti-proliferative action on the MDR human colon cancer and mouse lymphoma has been investigated.

Application

3-Formyl-6-methylchromone has been used for the synthesis of 3-formyl-6-methylchromone-4-phenylthiosemicarbazone.
3-Formyl-6-methylchromone may be used in the preparation of series of Schiff′s bases, via reaction with aromatic sulfonamides, such as sulfanilamide, homosulfanilamide, 4-aminoethyl-benzenesulfonamide, a pyrimidinyl-substituted sulfanilamide derivative, sulfaguanidine and 4-amino-6-trifluoromethyl-benzene-1,3-disulfonamide. It may be used in the preparation of Schiff base, which undergoes addition reaction with diethyl phosphite to afford bis[(1,2,4,3-triazaphospholyl)(chromonyl)aminomethylphosphonate] derivative and bis[(1,2,4,3-triazaphospholyl)(chromonyl)phosphorylamino-methylphosphonate] derivative.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis, characterization, crystal structure and antimicrobial activity of copper (II) complexes with a thiosemicarbazone derived from 3-formyl-6-methylchromone.
Ilies DC, et al.
Polyhedron, 81, 123-131 (2014)
Synthetic approach for novel bis (a-aminophosphonic acid) derivatives of chromone containing 1, 2, 4, 3-triazaphosphole moieties
Ali, TE and Halacheva, SS.
Heteroatom Chem., 20(3), 117-122 (2009)
Luca Puccetti et al.
Bioorganic & medicinal chemistry letters, 15(12), 3096-3101 (2005-05-24)
A series of Schiff's bases was prepared by reaction of 3-formyl-chromone or 6-methyl-3-formyl-chromone with aromatic sulfonamides, such as sulfanilamide, homosulfanilamide, 4-aminoethyl-benzenesulfonamide, a pyrimidinyl-substituted sulfanilamide derivative, sulfaguanidine and 4-amino-6-trifluoromethyl-benzene-1,3-disulfonamide. The zinc complexes of these sulfonamides have also been obtained. The new
Jessica Elizabeth Philip et al.
Journal of photochemistry and photobiology. B, Biology, 175, 178-191 (2017-09-12)
Two chromone hydrazone ligands HL
Enthalpies of combustion and formation of 3-formylchromones.
Flores H, et al.
Thermochimica Acta, 450(1), 35-37 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service