Skip to Content
Merck
All Photos(1)

Key Documents

38224

Sigma-Aldrich

Diisooctylthiophosphinic acid

technical, ~85% (T)

Synonym(s):

Bis(2,4,4-trimethylpentyl)thiophosphinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H35OPS
CAS Number:
Molecular Weight:
306.49
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
~85% (T)

grade

technical

Quality Level

Assay

~85% (T)

form

liquid

refractive index

n20/D 1.48

density

0.93 g/mL at 20 °C (lit.)

SMILES string

CC(CC(C)(C)C)CP(S)(=O)CC(C)CC(C)(C)C

InChI

1S/C16H35OPS/c1-13(9-15(3,4)5)11-18(17,19)12-14(2)10-16(6,7)8/h13-14H,9-12H2,1-8H3,(H,17,19)

InChI key

KUYLHALFMPOMKK-UHFFFAOYSA-N

General description

Diisooctylthiophosphinic acid (Bis(2,4,4-trimethylpentyl)thiophosphinic acid) is a hygroscopic liquid. Its physical properties (refractive index and specific gravity) have been reported.
Diisooctylthiophosphinic acid is an application specific reagent.

Application

Complexing agent for the extraction of transition metals
Diisooctylthiophosphinic acid is suitable for use in the colorimetric determination of airborne lead. It may be used as chelating agent for supercritical fluid extraction (SFE) of metals, for subsequent ICP-MS analyses of mercury, cadmium and lead in sediment.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raja Norimie Raja Sulaiman et al.
Journal of hazardous materials, 340, 77-84 (2017-07-18)
The discharge of electroplating waste containing nickel ions has led to environmental issues owing to the toxicity problem mainly to the aquatic organisms and humans. Liquid-liquid extraction offers a great potential treatment for nickel removal with several advantages of simple
Supercritical Fluid Extraction of Metals Using Bis (2, 4, 4-trimethyl-pentyl) monothiophosphinic Acid as Chelating Agent for Subsequent ICP-MS Analyses of Mercury, Cadmium and Lead in Sediment.
Roa EC, et al.
E-Journal of Chemistry, 8(3), 1114-1119 (2011)
Ash M and Ash I.
Handbook of solvents., 254-254 (2003)
Pornpimol Kongtip et al.
The Southeast Asian journal of tropical medicine and public health, 41(6), 1500-1511 (2011-02-19)
We developed a method to analyze airborne lead concentrations in the field. It was a modification of the colorimetric method using the reaction between 4(2-pyridylazo)-resorcinol (PAR) and lead with cyanex302 in an acid medium to reduce interfering metals. The lead

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service