Skip to Content
Merck
All Photos(3)

Key Documents

377449

Sigma-Aldrich

Manganese(II) carbonate

≥99.9% trace metals basis

Synonym(s):

Manganese carbonate, Manganese(2+) carbonate, Manganous carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnCO3
CAS Number:
Molecular Weight:
114.95
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder
solubility:
dilute aqueous acid: slightly soluble(lit.)

Quality Level

Assay

≥99.9% trace metals basis

form

powder

impurities

≤1,000.0 ppm Trace Metal Analysis

mp

>200 °C (lit.)

solubility

dilute aqueous acid: slightly soluble(lit.)

density

3.12 g/mL at 25 °C (lit.)

SMILES string

[Mn++].[O-]C([O-])=O

InChI

1S/CH2O3.Mn/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

XMWCXZJXESXBBY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Manganese(II) carbonate is a chemical compound that has a structure similar to calcite, with octahedral co-ordination symmetry. It is a carbonate that is insoluble in water and on treatment with acid it gives water soluble salts. It is a widely used material in plant fertilization as an additive that cures the magnesium deficiency in crops.

Application

Manganese(II) carbonate can be used as:      
  • A primary electrode material in asymmetric supercapacitors for improving the charge storage capacity and overall performance of the supercapacitors.      
  • A precursor for synthesizing manganese oxide, which is used as a component in various electrochemical applications, including batteries and supercapacitors.    
  • A precursor material in the synthesis of oxygen vacancy-rich nitrogen-doped manganese carbonate (MnCO2@N) microspheres. These microspheres are then employed as cathode materials in aqueous zinc-ion batteries (ZIBs) to enhance their electrochemical performance.
  • A potential electrocatalyst for the oxygen evolution reaction (OER) in water splitting applications.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structures of hydrothermally synthesized cobalt (II) carbonate and nickel (II) carbonate
Pertlik F
Acta Crystallographica Section E: Crystallographic Communications, 42(1), 4-5 (1986)
Jyoti Patel et al.
Nanomaterials (Basel, Switzerland), 10(5) (2020-05-24)
Norfloxacin (NOFX), a broadly used fluoroquinolone antibiotic, has been a subject of great concern in the past few years due to its undesirable effect on human beings and aquatic ecosystems. In this study, novel Mn doped ZnS (Mn:ZnS) quantum dots
K Maciejewska et al.
Nanomaterials (Basel, Switzerland), 10(3) (2020-03-04)
Herein, a novel synthesis method of colloidal GdPO4:Mn2+,Eu3+ nanoparticles for luminescent nanothermometry is proposed. XRD, TEM, DLS, and zeta potential measurements confirmed the crystallographic purity and reproducible morphology of the obtained nanoparticles. The spectroscopic properties of GdPO4:Mn2+,Eu3+ with different amounts
Manganese (II) salts as efficient catalysts for chemo selective transesterification of beta-keto esters under non-conventional conditions
Krishnaiah G, et al.
Tetrahedron Letters, 54(7), 703-706 (2013)
Manganese compounds
Reidies AH
Ullmann's Encyclopedia of Industrial Chemistry, 42(1), 4-5 (2000)

Articles

Recently, layer-by-layer (LbL) assembly has emerged as a versatile, gentle and, simple method for immobilization of functional molecules in an easily controllable thin film morphology.1,2 In this short review, we introduce recent advances in functional systems fabricated by using the mild, yet adaptable LbL technique.

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service