Skip to Content
Merck
All Photos(1)

Key Documents

376841

Sigma-Aldrich

3-Phenyl-1-propyne

contains ca.250 ppm BHT as inhibitor, 97%

Synonym(s):

1-Phenyl-2-propyne, 2-Propyn-1-ylbenzene, 2-Propynylbenzene, 3-Phenylpropyne, Benzylacetylene, Propargylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2C≡CH
CAS Number:
Molecular Weight:
116.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

contains

ca.250 ppm BHT as inhibitor

refractive index

n20/D 1.526 (lit.)

bp

75 °C/20 mmHg (lit.)

density

0.934 g/mL at 25 °C (lit.)

functional group

phenyl

SMILES string

C#CCc1ccccc1

InChI

1S/C9H8/c1-2-6-9-7-4-3-5-8-9/h1,3-5,7-8H,6H2

InChI key

NGKSKVYWPINGLI-UHFFFAOYSA-N

General description

3-Phenyl-1-propyne is a 3-aryl-1-propyne. The electronic transition of the resonance-stabilized 1-phenylpropargyl radical, produced in a jet-cooled discharge of 3-phenyl-1-propyne, has been studied by laser-induced fluorescence excitation and dispersed single vibronic level fluorescence (SVLF) spectroscopy. The microwave rotational spectrum of 3-phenyl-1-propyne (propargyl benzene) has been studied and its stable conformation is reported to have coplanar carbon atoms. Reaction of N-methyl-N-phenylhydrazine or N-phenylhydrazine with 3-phenyl-1-propyne is reported to yield indoles. 3-Phenyl-1-propyne is reported to react with styrene oxide and sodium azide, to afford β-hydroxytriazoles.

Application

3-Phenyl-1-propyne may be used as starting reagent in the synthesis of 4-phenyl-2-butyn-1-ol.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neil J Reilly et al.
The Journal of chemical physics, 130(14), 144313-144313 (2009-04-17)
The D(1)((2)A("))-D(0)((2)A(")) electronic transition of the resonance-stabilized 1-phenylpropargyl radical, produced in a jet-cooled discharge of 3-phenyl-1-propyne, has been investigated in detail by laser-induced fluorescence excitation and dispersed single vibronic level fluorescence (SVLF) spectroscopy.The transition is dominated by the origin band
Yadav JS, et al.
Tetrahedron Letters, 48(89), 8773-8776 (2007)
The rotational spectrum and heavy-atom-planar structure of propargyl benzene (3-phenyl-1-propyne).
Giudici R, et al.
Journal of Molecular Structure, 786(1), 65-67 (2006)
Yalin Zhang et al.
Bioorganic & medicinal chemistry, 12(14), 3847-3855 (2004-06-24)
The methionine salvage pathway allows the in vivo recovery of the methylthio moiety of methionine upon the formation of methylthioadenosine (MTA) from S-adenosylmethionine (SAM). The Fe(II)-containing form of acireductone dioxygenase (ARD) catalyzes the penultimate step in the pathway in Klebsiella
Zinc-promoted hydrohydrazination of terminal alkynes: an efficient domino synthesis of indoles.
Karolin Alex et al.
Angewandte Chemie (International ed. in English), 47(12), 2304-2307 (2008-02-12)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service