Skip to Content
Merck
All Photos(1)

Key Documents

375934

Sigma-Aldrich

N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide

≥95%, with 1% tert-Butyldimethylchlorosilane

Synonym(s):

MTBSTFA+TBDMSCl, 99:1, N-Methyl-N-tert-butyldimethylsilyltrifluoroacetamide, MTBSTFA + 1% TBDMSCl, Silylating mixture VI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18F3NOSi
CAS Number:
Molecular Weight:
241.33
Beilstein:
3606546
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Product Name

N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, ≥95%

Quality Level

Assay

≥95%

form

liquid

contains

1% TBDMSCl

functional group

amine
fluoro

storage temp.

−20°C

SMILES string

CN(C(=O)C(F)(F)F)[Si](C)(C)C(C)(C)C

InChI

1S/C9H18F3NOSi/c1-8(2,3)15(5,6)13(4)7(14)9(10,11)12/h1-6H3

InChI key

QRKUHYFDBWGLHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-butyldimethylchlorosilane is commonly used for the preparation of N-tert-butyldimethylsilyl ethanolamines resulting from the hydrolysis of nitrogen mustards. It is also used for selective O-silylation of N,O-diacylhydroxyl amines.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Organometallics, 23, 2157-2161 (2004)
Tsubasa Nakajima et al.
Plant & cell physiology, 55(9), 1605-1612 (2014-06-28)
Cyanobacteria have flexible metabolic capability that enables them to adapt to various environments. To investigate their underlying metabolic regulation mechanisms, we performed an integrated analysis of metabolic flux using transcriptomic and metabolomic data of a cyanobacterium Synechocystis sp. PCC 6803
P Canosa et al.
Journal of chromatography. A, 1072(1), 107-115 (2005-05-11)
A solid-phase microextraction (SPME) method for the determination of triclosan, methyl triclosan, 2,4-dichlorophenol and 2,3,4-trichlorophenol (considered as possible triclosan metabolites) in water samples was optimised. Analytes were first concentrated on a SPME fibre, directly exposed to the sample, and then
Joon Ho Kang et al.
Nature communications, 11(1), 4983-4983 (2020-10-07)
The energetic demands of a cell are believed to increase during mitosis, but the rates of ATP synthesis and consumption during mitosis have not been quantified. Here, we monitor mitochondrial membrane potential of single lymphocytic leukemia cells and demonstrate that
R Montes et al.
Journal of chromatography. A, 1216(2), 205-210 (2008-12-17)
A fast and novel sample preparation procedure for the determination of triclosan (TCS) and methyltriclosan (MTCS) in water samples is presented. Dispersive liquid-liquid microextraction, using a ternary mixture consisting of a disperser, an extractant and N-methyl-N-(tert-butyldimethylsilyl)trifluoroacetamide (MTBSTFA) as derivatization reagent

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service