Skip to Content
Merck
All Photos(1)

Key Documents

364460

Sigma-Aldrich

L-Proline benzyl ester hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO2 · HCl
CAS Number:
Molecular Weight:
241.71
Beilstein:
3598081
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D −48°, c = 1 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

148-151 °C (lit.)

application(s)

peptide synthesis

SMILES string

Cl[H].[H][C@]1(CCCN1)C(=O)OCc2ccccc2

InChI

1S/C12H15NO2.ClH/c14-12(11-7-4-8-13-11)15-9-10-5-2-1-3-6-10;/h1-3,5-6,11,13H,4,7-9H2;1H/t11-;/m0./s1

InChI key

NEDMOHHWRPHBAL-MERQFXBCSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Extensive applications in peptide chemistry and used as a chiral auxiliary in the asymmetric Diels-Alder reaction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantiomeric purity determination of L-proline benzyl ester by chiral column gas chromatography.
M Jemal et al.
Journal of chromatography, 392, 442-446 (1987-04-17)
A Córdova et al.
Bioorganic & medicinal chemistry letters, 9(21), 3119-3122 (1999-11-24)
Mesylates or tosylates of delta-hydroxy-L-norvaline esters spontaneously afford L-proline esters upon exposure to aqueous buffer in near quantitative yield. This novel reaction has led to the development of a simple route to optically active proline esters.
Yukihiro Yamamoto et al.
Applied and environmental microbiology, 76(18), 6180-6185 (2010-08-03)
We specifically examined an exopeptidase, prolyl aminopeptidase (PAP), as a target for synthesis of proline-containing peptides. A PAP from Streptomyces thermoluteus subsp. fuscus NBRC14270 (PAP14270) was obtained using sequence-based screening. From PAP14270, 144Ser was replaced by Cys (scPAP14270) to give
Aldrichimica Acta, 23, 45-45 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service