Skip to Content
Merck
All Photos(1)

Documents

357898

Sigma-Aldrich

Serinol

98%

Synonym(s):

2-Amino-1,3-propanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(NH2)CH2OH
CAS Number:
Molecular Weight:
91.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

277 °C (lit.)

mp

52-55 °C (lit.)

SMILES string

NC(CO)CO

InChI

1S/C3H9NO2/c4-3(1-5)2-6/h3,5-6H,1-2,4H2

InChI key

KJJPLEZQSCZCKE-UHFFFAOYSA-N

General description

Serinol activates toxin production in the attenuated cultures of Helminthosporium sacchari. Preparation of polyesters derived from serinol is reported.

Application

Serinol may be used to prepare the artificial nucleic acids. It may be used in the synthesis of N-acylated serinol and C16-serinol affinity gel.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

E Bieberich et al.
The Journal of biological chemistry, 275(1), 177-181 (2000-01-05)
A novel structural analog of ceramide was synthesized by N-acylation of serinol (2-amino-1,3-propanediol) and studied for its effects on glycolipid biosynthesis and cell differentiation of neuroblastoma cells. Incubation with N-palmitoylated serinol (C16-serinol) increased the concentration of endogenous ceramide by 50-80%
Gea Prioglio et al.
Polymers, 12(4) (2020-04-25)
The ultimate properties and resistance to fracture of nanocomposites based on poly(1,4-cis-isoprene) from Hevea Brasiliensis (natural rubber, NR) and a high surface area nanosized graphite (HSAG) were improved by using HSAG functionalized with 2-(2,5-dimethyl-1H-pyrrol-1-yl)propane-1,3-diol (serinol pyrrole) (HSAG-SP). The functionalization reaction
Hiroyuki Asanuma et al.
Chemical record (New York, N.Y.), 14(6), 1055-1069 (2014-08-30)
In this account, we demonstrate a new methodology for the de novo design of functional oligonucleotides with the acyclic scaffolds threoninol and serinol. Four functional motifs-wedge, interstrand-wedge, dimer, and cluster-have been prepared from natural DNA or RNA and functional base
Daniele Locatelli et al.
Nanomaterials (Basel, Switzerland), 10(6) (2020-06-21)
The solubility parameters of multiwalled carbon nanotubes (CNTs) was tuned via their chemical modification with pyrrole compounds (PyCs), by means of a simple and sustainable methodology. PyCs were synthesized with high atom efficiency through the Paal-Knorr reaction of primary amines
Control of the chirality and helicity of oligomers of serinol nucleic acid (SNA) by sequence design.
Hiromu Kashida et al.
Angewandte Chemie (International ed. in English), 50(6), 1285-1288 (2011-02-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service