If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf
357391
Silicon carbide
−400 mesh particle size, ≥97.5%
Synonym(s):
Carbon silicide, Carborundum, Methanidylidynesilanylium, Silicon monocarbide
About This Item
Recommended Products
description
hexagonal phase
Quality Level
Assay
≥97.5%
form
powder
particle size
−400 mesh
mp
2700 °C (lit.)
density
3.22 g/mL at 25 °C (lit.)
SMILES string
[C-]#[Si+]
InChI
1S/CSi/c1-2
InChI key
HBMJWWWQQXIZIP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Three approaches generate white light, including LED-based down-conversion for broader applications.
Three approaches generate white light, including LED-based down-conversion for broader applications.
Three approaches generate white light, including LED-based down-conversion for broader applications.
Three approaches generate white light, including LED-based down-conversion for broader applications.
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
-
What is the specific surface area of this product?
1 answer-
The surface area is approximately 0.25 m2/g, but it is not measured on every lot.
Helpful?
-
-
こちらの製品は 労安規則(577条の2)がん原性物質 でしょうか
1 answer-
Classifications set by the Labor Safety Regulations are out of scope and not reported in the current SDS document. The Carcinogenicity values offered by other agencies may be found in Sections 8 and 11 of the SDS. Please see the link below to review this document:
https://www.sigmaaldrich.com/sds/aldrich/357391Helpful?
-
-
Where can I find other Silicon- or Carbon-based materials?
1 answer-
Sigma-Aldrich offers a wide variety of Silicon-, Carbon-, and other metal-based ceramic materials.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
Is Silicon carbide, Product 357391, alpha- or beta-phase?
1 answer-
According to our supplier, this product is alpha-phase with a hexagonal crystal structure.
Helpful?
-
-
What solvents can I use to dissolve Silicon carbide, Product 357391?
1 answer-
Unfortunately, Sigma-Aldrich does not test the solubility of this material. Fundamentally, Silicon carbide (SiC) is insoluble in water. However, according to Lange's Handbook of Chemistry (16th ed.), beta-phase SiC is expected to be soluble in fused alkali oxides (e.g. KOH) at a ratio of 1 part per 100 parts. No specific solubility information is available for this alpha-phase product.
Helpful?
-
-
What applications can Silicon carbide, Product 357391, be used for?
1 answer-
Silicon carbide (SiC) has found use in a variety of commercial applications. Historically, SiC has been used most frequently in the metallurical, abrasive, and refactory industries. More recently, this material has found increased use in high technology applications (e.g. electronics, aerospace, automotive, etc.).
Helpful?
-
-
How does the storage temperature relate to shipping conditions?
1 answer-
The storage conditions that a Sigma-Aldrich catalog and label recommend for products are deliberately conservative. For many products, long-term storage at low temperatures will increase the time during which they are expected to remain in specification and therefore are labeled accordingly. Where short-term storage, shipping time frame, or exposure to conditions other than those recommended for long-term storage will not affect product quality, Sigma-Aldrich will ship at ambient temperature. The products sensitive to short-term exposure to conditions other than their recommended long-term storage are shipped on wet or dry ice. Ambient temperature shipping helps to control shipping costs for our customers. At any time, our customers can request wet- or dry-ice shipment, but the special handling is at customer expense if our product history indicates that the product is stable for regular shipment.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service