Skip to Content
Merck
All Photos(3)

Key Documents

346713

Sigma-Aldrich

(1S,2S)-(+)-1,2-Diaminocyclohexane

98%

Synonym(s):

(1S)-trans-1,2-Cyclohexanediamine, (1S)-trans-1,2-Diaminocyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(NH2)2
CAS Number:
Molecular Weight:
114.19
Beilstein:
2801645
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]20/D +25°, c = 5 in 1 M HCl

optical purity

ee: 99% (GLC)

bp

104-110 °C/40 mmHg (lit.)

mp

40-43 °C (lit.)

SMILES string

N[C@H]1CCCC[C@@H]1N

InChI

1S/C6H14N2/c7-5-3-1-2-4-6(5)8/h5-6H,1-4,7-8H2/t5-,6-/m0/s1

InChI key

SSJXIUAHEKJCMH-WDSKDSINSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Versatile ligand for the formation of metal complexes. Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luka Ðorđević et al.
Nature communications, 9(1), 3442-3442 (2018-08-26)
The chirality of (nano)structures is paramount in many phenomena, including biological processes, self-assembly, enantioselective reactions, and light or electron spin polarization. In the quest for new chiral materials, metallo-organic hybrids have been attractive candidates for exploiting the aforementioned scientific fields.
Mukaiyama, T.
Aldrichimica Acta, 29, 59-59 (1996)
Chenier, P.J. et al.
Tetrahedron Letters, 38, 7341-7341 (1997)
Hidekazu Yamada et al.
Journal of the American Chemical Society, 134(17), 7250-7253 (2012-04-18)
Optically active amidine dimer strands having a variety of chiral and achiral linkers with different stereostructures are synthesized and used as templates for diastereoselective imine-bond formations between two achiral carboxylic acid monomers bearing a terminal aldehyde group and racemic 1,2-cyclohexanediamine
Ran Feng et al.
Dalton transactions (Cambridge, England : 2003), 42(6), 2130-2145 (2012-11-29)
In this paper, the origins of enantioselectivity in asymmetric ketone hydrogenation catalyzed by RuH(2)(binap)(cydn) (cydn = trans-1,2-diaminocyclohexane) were discussed. Fifteen substrates involving aromatic, heteroaromatic, olefinic and dialkyl prochiral ketones were used to probe the catalytic mechanism and find an effective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service