Skip to Content
Merck
All Photos(2)

Key Documents

344443

Sigma-Aldrich

Sodium fluorophosphate

95%

Synonym(s):

Disodium fluorophosphate, Disodium monofluorophosphate, Disodium phosphorofluoridate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2FPO3
CAS Number:
Molecular Weight:
143.95
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
95%
form:
powder
solubility:
H2O: slightly soluble(lit.)

Quality Level

Assay

95%

form

powder

solubility

H2O: slightly soluble(lit.)

SMILES string

[Na+].[Na+].[O-]P([O-])(F)=O

InChI

1S/FH2O3P.2Na/c1-5(2,3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

BFDWBSRJQZPEEB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium fluorophosphate is a white crystalline compound that is slightly soluble in water. It is used to fabricate cathode material for Na-ion batteries and also acts as a corrosion inhibitor. It also finds application in the field of oral care products and specialty glasses.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles Parkinson et al.
American journal of dentistry, 26 Spec No A, 25A-31A (2013-07-10)
To compare the efficacy of a 0.454% w/w stannous fluoride containing anhydrous dentifrice and a negative control dentifrice containing 1,000 ppm fluoride (as sodium monofluorophosphate) at reducing dentin hypersensitivity over an 8-week period, following twice daily brushing. This was a
Yiming Li et al.
American journal of dentistry, 25(1), 21-25 (2012-05-09)
To investigate whether the long-term use (6 months) of an arginine-calcium carbonate-MFP toothpaste would affect calculus formation and/or gingivitis when compared to a calcium carbonate-MFP toothpaste. This was a double-blind clinical study. Eligible adult subjects (120) entered a 2-month pre-test
Aline Silva Braga et al.
Biofouling, 34(3), 252-261 (2018-02-13)
The purpose of this study was to evaluate the antimicrobial (anti-biofilm) and anti-caries (enamel demineralization prevention) effects of Malva sylvestris (Malvatricin® Plus) compared with known antimicrobial mouthrinses. Microcosm biofilm was produced on enamel, using inoculum from pooled human saliva mixed
David Hamlin et al.
American journal of dentistry, 25(3), 146-152 (2012-09-20)
Dentin hypersensitivity is a significant clinical problem that affects numerous individuals. This sharp pain, arising from exposed dentin in response to external stimuli, can be a particularly uncomfortable and unpleasant sensation for patients, because it interferes with their quality of
M C Aguirre et al.
Mathematical biosciences, 234(1), 17-24 (2011-08-26)
Alpha-macroglobulins (AM) are proteins that inactivate proteinases. Sodium monofluorophosphate (MFP) binds to AM and transiently changes AM plasma levels. As a consequence MFP is useful to modify AM homeostasis. A mathematical model to study the homeostasis of AM is proposed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service