Skip to Content
Merck
All Photos(1)

Key Documents

343455

Sigma-Aldrich

2-Methoxytetrahydropyran

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O2
CAS Number:
Molecular Weight:
116.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.425 (lit.)

bp

128-129 °C (lit.)

density

0.963 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

COC1CCCCO1

InChI

1S/C6H12O2/c1-7-6-4-2-3-5-8-6/h6H,2-5H2,1H3

InChI key

XTDKZSUYCXHXJM-UHFFFAOYSA-N

General description

2-Methoxytetrahydropyran is methoxy-substituted tetrahydropyran. α- and β-glycosidic C1-O1 bonds of the axial and equatorial forms of 2-methoxytetrahydropyran has been investigated by ab initio conformational study. Stereochemical properties of the glycosidic linkage of 2-methoxytetrahydropyran have been studied by the quantum-chemical PCILO method. It undergoes elimination reaction in the gas phase to yield 3, 4-dihydro-2H-pyran and methanol. Proton resonance spectra of 2-methoxytetrahydropyran has been analyzed.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proton chemical shifts in NMR. Part 12. 1 Steric, electric field and conformational effects in acyclic and cyclic ethers.
Abraham RJ and Warne MA.
J. Chem. Soc. Perkin Trans. II, 8, 1751-1758 (1998)
The conformational properties of the glycosidic linkage.
Tvaroska I and Kozar T.
Carbohydrate Research, 90(2), 173-185 (1981)
Combined experimental and theoretical studies of the elimination kinetic of 2-methoxytetrahydropyran in the gas phase.
Rosas F, et al.
Journal of Physical Organic Chemistry, 23(12), 1127-1136 (2010)
Rodrigo S Bitzer et al.
Carbohydrate research, 340(13), 2171-2184 (2005-08-02)
An ab initio conformational study of the alpha- and beta-glycosidic C1-O1 bonds has been carried out on the axial and equatorial forms of 2-methoxytetrahydropyran (2-MTHP) at the HF/6-31G(d,p) and GVB-PP/6-31G(d,p) levels of calculation. Six conformers of 2-MTHP were fully optimized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service