Skip to Content
Merck
All Photos(1)

Key Documents

343412

Sigma-Aldrich

Cyclopropane-1,1-dicarboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H4(CO2H)2
CAS Number:
Molecular Weight:
130.10
Beilstein:
1864823
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

134-136 °C (lit.)

solubility

methanol: soluble 1 g/10 mL, clear, colorless

functional group

carboxylic acid

SMILES string

OC(=O)C1(CC1)C(O)=O

InChI

1S/C5H6O4/c6-3(7)5(1-2-5)4(8)9/h1-2H2,(H,6,7)(H,8,9)

InChI key

FDKLLWKMYAMLIF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclopropane-1,1-dicarboxylic acid is a dicarboxylic acid. Cyclopropane-1,1-dicarboxylic acid, an inhibitor of 1-aminocyclopropane-1-carboxylic acid oxidase, was quantitated in Lycopersicum esculentum by HPLC-electrospray tandem mass spectrometry. Crystal and molecular structure of cyclopropane-1,1-dicarboxylic acid has been reported.

Application

Cyclopropane-1,1-dicarboxylic acid was used in the preparation of new heterocyclic derivatives of cyclopropane dicarboxylic acid containing thiadiazole and 1,2,4-triazole moieties. It was also used to prepare spiro-cyclopropyl metallocycles.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The crystal and molecular structure of cyclopropane-1, 1-dicarboxylic acid.
Meester MAM, et al.
Acta Crystallographica Section B, Structural Science, 27(3), 630-634 (1971)
Tetrahedron, 45, 1219-1219 (1989)
Y H Dong et al.
Science in China. Series B, Chemistry, life sciences & earth sciences, 33(11), 1297-1303 (1990-11-01)
The crystal structures of [Pt(NH3)2CPrDCA].H2O (I), [Pt(CH3NH2)2CPrDCA] (II), and [Pt(dmbn) CPrDCA].2.5H2O (III) (where CPrDCA is 1,1-cyclopropanedicarboxylate; dmbn is 2,3-dimethyl-2,3-butyldiamine) are determined. Compound I crystallizes in the orthorhombic space group Pnma with the cell dimensions: a = 6.517(2), b = 9.709(3)
A Hussain K Sharba et al.
Molecules (Basel, Switzerland), 10(9), 1153-1160 (2007-11-17)
New heterocyclic derivatives of cyclopropane dicarboxylic acid comprising thiadiazole and 1,2,4-triazole moieties are reported. Reaction of 1,1-cyclopropane dicarboxylic acid (1) with thiosemicarbazide and phosphorous oxychloride resulted in 1,1-bis (2-amino-1,3,4-thiadiazol-5- yl)cyclopropane (2). Cyclopropane dicarboxylic acid thiosemicarbazide (6) was converted into 1,1-bis(3-thio-4H-1,2,4-triazol-5-yl)
Michael B Johansen et al.
Organic letters, 10(16), 3497-3500 (2008-07-12)
Pyrrolo[1,2- a]indoles are conveniently prepared from tetrahydro-1,2-oxazines, which in turn are generated through the reaction of nitrones with 1,1-cyclopropanediesters. The synthetic route proves to be highly diastereoselective and provides access to the core of the recently discovered pyrrolo[1,2- a]indole natural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service