Skip to Content
Merck
All Photos(1)

Key Documents

34125

Sigma-Aldrich

Dibromodifluoromethane

≥95.0% (GC)

Synonym(s):

Carbon dibromide difluoride, Difluorodibromomethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CBr2F2
CAS Number:
Molecular Weight:
209.82
Beilstein:
1732515
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

7.24 (vs air)

vapor pressure

12.79 psi ( 20 °C)

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.398 (lit.)
n20/D 1.398

bp

22-23 °C (lit.)

mp

−142-−141 °C (lit.)

density

2.297 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(Br)Br

InChI

1S/CBr2F2/c2-1(3,4)5

InChI key

AZSZCFSOHXEJQE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The photolysis products of dibromodifluoromethane were characterized by matrix isolation infrared and UV/Visible spectroscopy. Free radical addition of dibromodifluoromethane with fluoroolefins has been reported. Dibromodifluoromethane reacts with hydrocarbon olefins to yield 1,3-dibromo-1,1-difluoroalkene. Sulfinatodehalogenation reagent promoted addition reaction of difluorodibromomethane with alkenes, alkynes and cyclic enol ethers has been described.

Application

  • Synthesis of 3‑Fluoropyrazoles via Radical-Polar Crossover Photoinduced Cyclization: This study highlights the use of dibromodifluoromethane (CF2Br2) for the synthesis of valuable difluorinated and monofluorinated compounds through a novel photoinduced cyclization process (MA Reed, ND Patil, Synfacts, 2023).

Other Notes

Reagent for the Wittig olefination of carbonyl compounds to difluoromethylene and fluoromethylene derivatives; Precursor of difluorocarbene

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ozone 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H.P. Fritz et al.
Zeitschrift fur Naturforschung, B: Chemical Sciences, 86, 1375-1375 (1981)
Free Radical Additions Involving Fluorine Compounds. IV. The Addition of Dibromodifluoromethane to Some Fluoroolefins1.
Tarrant P, et al.
Journal of the American Chemical Society, 77(10), 2783-2787 (1955)
Free Radical Additions Involving Fluorine Compounds. I. The Addition of Dibromodifluoromethane to Hydrocarbon Olefins.
Tarrant P and Lovelace AM.
Journal of the American Chemical Society, 76(13), 3466-3468 (1954)
Addition reactions of dibromodifluoromethane promoted by sulfinatodehalogenation reagents.
Wu F-H, et al.
Journal of Fluorine Chemistry, 80(2), 91-94 (1996)
S. Hayaski et al.
Chemistry Letters (Jpn), 983-983 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service