Skip to Content
Merck
All Photos(1)

Documents

337250

Sigma-Aldrich

4-Fluoro-3-methylphenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CH3)OH
CAS Number:
Molecular Weight:
126.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.515 (lit.)

bp

76 °C/5 mmHg (lit.)

mp

32 °C (lit.)

density

1.134 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(O)ccc1F

InChI

1S/C7H7FO/c1-5-4-6(9)2-3-7(5)8/h2-4,9H,1H3

InChI key

RVYGYYVGWSCWGY-UHFFFAOYSA-N

Application

4-Fluoro-3-methylphenol was used to detect the aromatic metabolites in methanogenic m-cresol-degrading consortium. It was also used in the preparation of 8-fluoronaphthoquinone via Friedel Crafts acylation reaction with maleic anhydride.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K L Londry et al.
Applied and environmental microbiology, 59(7), 2229-2238 (1993-07-01)
Anaerobic sewage sludge was used to enrich a methanogenic m-cresol-degrading consortium. 6-Fluoro-3-methylphenol was synthesized and added to subcultures of the consortium with m-cresol. This caused the accumulation of 4-hydroxy-2-methylbenzoic acid. In a separate experiment, the addition of 3-fluorobenzoic acid caused
Anita Mahapatra et al.
Bioorganic & medicinal chemistry, 15(24), 7638-7646 (2007-09-25)
The naphthoquinone 7-methyljuglone (5-hydroxy-7-methyl-1,4-naphthoquinone) has previously been isolated and identified as an active component of root extracts of Euclea natalensis which displays antitubercular activity. Herein, a series of synthetic and plant-derived naphthoquinone derivates of the 7-methyljuglone scaffold have been prepared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service