Skip to Content
Merck
All Photos(1)

Key Documents

33040

Sigma-Aldrich

2,5-Diaminohydroquinone dihydrochloride

technical, ≥90% (AT)

Synonym(s):

2,5-Diamino-1,4-benzenediol dihydrochloride, 2,5-Diamino-1,4-dihydroxybenzene dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O2 · 2HCl
CAS Number:
Molecular Weight:
213.06
Beilstein:
5795549
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (AT)

form

powder

SMILES string

Cl.Cl.Nc1cc(O)c(N)cc1O

InChI

1S/C6H8N2O2.2ClH/c7-3-1-5(9)4(8)2-6(3)10;;/h1-2,9-10H,7-8H2;2*1H

InChI key

NILKAWPWTYPHAH-UHFFFAOYSA-N

Application

2,5-Diaminohydroquinone dihydrochloride was used in the preparation of polymers. It was also used in the preparation of chemically cross-linked polyimide and silica hybrid films.

Caution

may discolor to black-violet on storage

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Louis Sieuw et al.
Chemical science, 10(2), 418-426 (2019-02-13)
Small organic materials are generally plagued by their high solubility in battery electrolytes. Finding approaches to suppress solubilization while not penalizing gravimetric capacity remains a challenge. Here we propose the concept of a hydrogen bond stabilized organic battery framework as
Synthesis and characterization of chemically cross-linked polyimide-siloxane hybrid films.
Ahmad Z, et al.
Journal of Non-Crystalline Solids, 355(8), 507-517 (2009)
Polymers derived from 2, 5-diamino-p-benzoquinone-diimide and related compounds.
Szita J and Marvel CS.
Journal of Polymer Science Part A: Polymer Chemistry, 7(11), 3203-3217 (1969)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service