Skip to Content
Merck
All Photos(3)

Documents

325996

Sigma-Aldrich

Thulium(III) nitrate pentahydrate

powder or crystals, 99.9%

Synonym(s):

Thulium trinitrate pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Tm(NO3)3 · 5H2O
CAS Number:
Molecular Weight:
445.03
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

99.9%

form

powder or crystals

reaction suitability

core: thulium
reagent type: catalyst

SMILES string

O.O.O.O.O.[Tm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.5H2O.Tm/c3*2-1(3)4;;;;;;/h;;;5*1H2;/q3*-1;;;;;;+3

InChI key

PEYIBPJKEYRLDB-UHFFFAOYSA-N

Application

Thulium(III) nitrate pentahydrate has been used in the synthesis of thulium oxide (Tm2O3) nanorods for modified electrode in enzyme-based biosensor analysis.

It can also be used to synthesize hydrophilic, microporous lanthanide-organic frameworks.

Pictograms

Flame over circle

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A dual enzyme functionalized nanostructured thulium oxide based interface for biomedical application.
Singh J, et al.
Nanoscale, 6(2), 1195-1208 (2014)
Albenc Nexha et al.
Nanomaterials (Basel, Switzerland), 11(2) (2021-03-07)
The incorporation of oleic acid and oleylamine, acting as organic surfactant coatings for a novel solvothermal synthesis procedure, resulted in the formation of monoclinic KLu(WO4)2 nanocrystals. The formation of this crystalline phase was confirmed structurally from X-ray powder diffraction patterns
Hydrophilic microporous lanthanide-organic frameworks based on 4, 4?-biphenyldiacetate: Synthesis, crystal structures and sorption properties.
Makhloufi G, et al.
Polyhedron, 127, 59-67 (2017)
Manoj Kumar et al.
Cancer biotherapy & radiopharmaceuticals, 34(1), 24-32 (2018-10-03)
This article describes the preparation of a 170Tm source by chemical deposition technique, its encapsulation in a titanium holder, and preliminary quality evaluation for potential utility as a brachytherapy source. The procedure consisted of electrodeposition of Ni on a Cu

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service