323144
(S)-(−)-α−Methyl-2-naphthalenemethanol
98%
Synonym(s):
(S)-(−)-1-(2-Naphthyl)ethanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C10H7CH(CH3)OH
CAS Number:
Molecular Weight:
172.22
Beilstein:
2042396
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
optical activity
[α]20/D −40°, c = 5 in ethanol
mp
70-71 °C (lit.)
functional group
hydroxyl
SMILES string
C[C@H](O)c1ccc2ccccc2c1
InChI
1S/C12H12O/c1-9(13)11-7-6-10-4-2-3-5-12(10)8-11/h2-9,13H,1H3/t9-/m0/s1
InChI key
AXRKCRWZRKETCK-VIFPVBQESA-N
Gene Information
human ... UGT2B17(7367) , UGT2B7(7364)
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ingo Bichlmaier et al.
Journal of medicinal chemistry, 49(5), 1818-1827 (2006-03-03)
A set of 28 enantiomers comprising rigid and flexible secondary alcohols was synthesized by the asymmetric Corey-Bakshi-Shibata reduction. The enantiomerically pure alcohols were subjected to enzymatic glucuronidation assays employing the human UDP-glucuronosyltransferases (UGTs) 2B7 and 2B17. Both UGTs displayed high
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service