Skip to Content
Merck
All Photos(2)

Key Documents

308463

Sigma-Aldrich

Telluric acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H6TeO6
CAS Number:
Molecular Weight:
229.64
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

97.5-102.5%
98%

SMILES string

O[Te](O)(O)(O)(O)O

InChI

1S/H6O6Te/c1-7(2,3,4,5)6/h1-6H

InChI key

FXADMRZICBQPQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Telluric acids exist in both cubic and monoclinic crystalline forms.1 It may be prepared by treating tellurium and tellurous acid with strong oxidants.

Packaging

Packaged in glass bottles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wiberg E and Wiberg N
Inorganic Chemistry null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service