Skip to Content
Merck
All Photos(3)

Documents

306339

Sigma-Aldrich

Lanthanum(III) acetate hydrate

99.9% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
La(CH3CO2)3 · xH2O
CAS Number:
Molecular Weight:
316.04 (anhydrous basis)
Beilstein:
3693604
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace rare earth metals basis

form

crystalline powder

composition

Degree of hydration, ≤2

reaction suitability

core: lanthanum
reagent type: catalyst

SMILES string

O.CC(=O)O[La](OC(C)=O)OC(C)=O

InChI

1S/3C2H4O2.La.H2O/c3*1-2(3)4;;/h3*1H3,(H,3,4);;1H2/q;;;+3;/p-3

InChI key

YRKCZRMEPGLHRN-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Used in the synthesis of a novel phenolate-bridged dilanthanum(III) complex which has potential as a model for metalloproteins as well as in basic and applied chemistry.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wang, Z. et al.
Inorganic Chemistry, 36, 629-629 (1997)
Bo Zhou et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 5(3), 1700667-1700667 (2018-03-30)
Upconverting materials have achieved great progress in recent years, however, it remains challenging for the mechanistic research on new upconversion strategy of lanthanides. Here, a novel and efficient strategy to realize photon upconversion from more lanthanides and fine control of

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service