Skip to Content
Merck
All Photos(1)

Documents

304476

Sigma-Aldrich

5-Fluoro-2-methylbenzothiazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6FNS
CAS Number:
Molecular Weight:
167.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.584 (lit.)

bp

110-112 °C/7 mmHg (lit.)

density

1.256 g/mL at 25 °C (lit.)

SMILES string

Cc1nc2cc(F)ccc2s1

InChI

1S/C8H6FNS/c1-5-10-7-4-6(9)2-3-8(7)11-5/h2-4H,1H3

InChI key

LDBFGRGBYVULTJ-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuseppe Manfroni et al.
Bioorganic & medicinal chemistry, 20(2), 866-876 (2011-12-27)
Hepatitis C virus (HCV) infection has been recognized as the major cause of liver failure that can lead to hepatocellular carcinoma. Among all the HCV proteins, NS5B polymerase represents a leading target for drug discovery strategies. Herein, we describe our

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service