Skip to Content
Merck
All Photos(1)

Key Documents

299847

Sigma-Aldrich

2-Hydroxyfluorene

98%

Synonym(s):

2-Fluorenol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10O
CAS Number:
Molecular Weight:
182.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

167-169 °C (lit.)

SMILES string

Oc1ccc-2c(Cc3ccccc-23)c1

InChI

1S/C13H10O/c14-11-5-6-13-10(8-11)7-9-3-1-2-4-12(9)13/h1-6,8,14H,7H2

InChI key

ZDOIAPGLORMKTR-UHFFFAOYSA-N

General description

Urinary detection of 2-hydroxyfluorene in smokers and nonsmokers by HPLC with fluorescence detection was reported. 2-Hydroxyfluorene forms 1:1 complexes with β-cyclodextrin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

β-Cyclodextrin inclusion complexes of 2-hydroxyfluorene and 2-hydroxy-9-fluorenone: differences in stoichiometry and excited state prototropic equilibrium.
Enoch IVMV and Yousuf S.
Journal of Solution Chemistry, 42(2), 470-484 (2013)
Urinary 2-hydroxyfluorene and 1-hydroxypyrene levels in smokers and nonsmokers in Japan and Thailand.
Chetiyanukornkul T, et al.
Polycyclic Aromat. Compd., 24(4-5), 467-474 (2004)
Salwa Hajir et al.
Environmental pollution (Barking, Essex : 1987), 272, 115967-115967 (2020-11-13)
In Lebanon, previous studies have indicated an onset of cardiovascular diseases 12 years earlier than in other parts of the world, suggesting the presence of additional risk factors specific to Lebanon. Measurements of airborne particles in Lebanon surpass the recommendations
Pan Yang et al.
Environmental science & technology, 51(2), 958-967 (2016-12-15)
Toxicological studies have demonstrated that polycyclic aromatic hydrocarbons (PAHs) exposure impairs male reproductive health. However, the epidemiological evidence is limited and discordant. Our goal was to investigate the relationship between PAH exposures and human semen quality. We analyzed 12 urinary
Xiang-Yin Lou et al.
Environmental science and pollution research international, 26(26), 27281-27290 (2019-07-22)
Exposure to polycyclic aromatic hydrocarbons (PAHs) during pregnancy may pose adverse health risk to both the mothers and babies. In the present study, 188 pregnant women of different trimesters were recruited in Guangzhou, south China, and nine hydroxyl PAHs (OH-PAHs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service