Skip to Content
Merck
All Photos(2)

Key Documents

299030

Sigma-Aldrich

Methyl decanoate

99%

Synonym(s):

Capric acid methyl ester, Decanoic acid methyl ester, Methyl caprate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8COOCH3
CAS Number:
Molecular Weight:
186.29
Beilstein:
1759170
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

liquid

refractive index

n20/D 1.425 (lit.)

bp

108 °C/10 mmHg
224 °C (lit.)

mp

−14-−11 °C (lit.)

solubility

chloroform: soluble 100 mg/mL, clear

density

0.871 g/mL at 20 °C (lit.)

functional group

ester

SMILES string

CCCCCCCCCC(=O)OC

InChI

1S/C11H22O2/c1-3-4-5-6-7-8-9-10-11(12)13-2/h3-10H2,1-2H3

InChI key

YRHYCMZPEVDGFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thermal decomposition of methyl decanoate in a jet-stirred reactor at 773 to 1123K temperatures was studied[1]. The oxidation of methyl decanoate in a jet-stirred reactor at temperatures from 500 to 1100K was studied[2].

Application


  • Application of mist to fingermark detection: Misting with high-boiling-point liquid containing p-dimethylaminocinnamaldehyde and cyanoacrylate.: This article explored the use of high-boiling-point liquids, including methyl decanoate, in misting techniques for forensic fingermark detection. (Takatsu et al., 2023).

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

230.9 °F - Pensky-Martens closed cup

Flash Point(C)

110.5 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre Alexandre Glaude et al.
Combustion and flame, 157(11), 2035-2050 (2010-11-01)
The modeling of the oxidation of methyl esters was investigated and the specific chemistry, which is due to the presence of the ester group in this class of molecules, is described. New reactions and rate parameters were defined and included
Olivier Herbinet et al.
Combustion and flame, 158(7), 1288-1300 (2011-07-01)
The experimental study of the thermal decomposition of methyl decanoate was performed in a jet-stirred reactor at temperatures ranging from 773 to 1123 K, at residence times between 1 and 4 s, at a pressure of 800 Torr (106.6 kPa)
Hanneke Brust et al.
Journal of chromatography. A, 1374, 224-230 (2014-12-10)
In this work, a reliable and robust vacuum-outlet gas chromatography-mass spectrometry (GC-MS) method is introduced for the identification and quantification of impurities in trinitrotoluene (TNT). Vacuum-outlet GC-MS allows for short analysis times; the analysis of impurities in TNT was performed
Maria Makarova et al.
Current biology : CB, 30(3), 367-380 (2020-01-21)
Membrane function is fundamental to life. Each species explores membrane lipid diversity within a genetically predefined range of possibilities. How membrane lipid composition in turn defines the functional space available for evolution of membrane-centered processes remains largely unknown. We address
Oier Aizpurua-Olaizola et al.
Journal of food science, 80(1), E101-E107 (2014-12-05)
In this study, supercritical fluid extraction has been successfully applied to a sequential fractionation of fatty acids and polyphenols from wine wastes (2 different vitis vinifera grapes). To this aim, in a 1st step just fatty acids were extracted and

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service