Skip to Content
Merck
All Photos(1)

Key Documents

294632

Sigma-Aldrich

Methyl dimethoxyacetate

97%

Synonym(s):

Glyoxylic acid methyl ester dimethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2CHCOOCH3
CAS Number:
Molecular Weight:
134.13
Beilstein:
1757582
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.405 (lit.)

bp

67 °C/18 mmHg (lit.)

density

1.096 g/mL at 25 °C (lit.)

functional group

acetal
ester
ether

storage temp.

2-8°C

SMILES string

COC(OC)C(=O)OC

InChI

1S/C5H10O4/c1-7-4(6)5(8-2)9-3/h5H,1-3H3

InChI key

NZTCVGHPDWAALP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl dimethoxyacetate has been used:
  • in Claisen acylation of active hydrogen compounds
  • in preparation of 3, 9-disubstituted 2,4,8,10-tetroxaspiro [5.5] undecane
  • as Lithium enolate precursor
  • as acylating reagent for cycloalkanone enolates and amino alcohols

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron, 46, 6799-6799 (1990)
P J Connolly et al.
Journal of medicinal chemistry, 36(23), 3674-3685 (1993-11-12)
Compounds comprising a series of 7-[2-(4-fluorophenyl)-4,5,6,7-tetrahydro-2H-indazol-3-yl]-3,5- dihydroxy-6-heptenoic acid sodium salts (18) were synthesized and tested for their ability to inhibit HMG-CoA reductase in a partially purified enzyme preparation and cholesterol biosynthesis from acetate in cultured HEP-G2 cells. Changing the size
Synthetic Communications, 23, 1003-1003 (1993)
Studies in Mixed Ester Condensations. IV. Acylations with Methyl Dimethoxyacetate1.
Royals EE and Robinson AG.
Journal of the American Chemical Society, 78(16), 4161-4164 (1956)
Some 3, 9-Dicarboxylic Acids of 2, 4, 8, 10-Tetroxaspiro [5.5] undecane.
CLEMENTS JB and RICE LM.
The Journal of Organic Chemistry, 24(12), 1958-1961 (1959)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service