Skip to Content
Merck
All Photos(3)

Key Documents

289868

Sigma-Aldrich

Methylphosphonic acid

98%

Synonym(s):

MPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3P(O)(OH)2
CAS Number:
Molecular Weight:
96.02
Beilstein:
1739372
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

105-107 °C (lit.)

SMILES string

CP(O)(O)=O

InChI

1S/CH5O3P/c1-5(2,3)4/h1H3,(H2,2,3,4)

InChI key

YACKEPLHDIMKIO-UHFFFAOYSA-N

Gene Information

human ... ALPP(250)

Looking for similar products? Visit Product Comparison Guide

General description

The vibrational spectra of aqueous solutions of methylphosphonic acid and its anions were studied. The supercritical water co-oxidation elementary reaction rate mechanism for methylphosphonic acid was also studied.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>392.0 °F - Pensky-Martens closed cup

Flash Point(C)

> 200 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vibrational analysis of methylphosphonic acid and its anions: I. Vibrational spectra.
Journal of Molecular Structure, 15(2), 225-236 (1973)
Co-oxidation of methylphosphonic acid and ethanol in supercritical water: II: Elementary reaction rate model.
Journal of Supercritical Fluids, 39(2), 239-245 (2006)
Janel Owens et al.
Journal of agricultural and food chemistry, 57(18), 8227-8235 (2009-08-19)
Though chemical warfare agents (CWAs) have been banned by the Chemical Weapons Convention, the threat that such chemicals may be used, including their deliberate addition to food, remains. In such matrixes, CWAs may hydrolyze to phosphonic acids, which are good
Chien-Hui Ma et al.
The EMBO journal, 28(12), 1745-1756 (2009-05-15)
Water, acting as a rogue nucleophile, can disrupt transesterification steps of important phosphoryl transfer reactions in DNA and RNA. We have unveiled this risk, and identified safeguards instituted against it, during strand cleavage and joining by the tyrosine site-specific recombinase
James Pratt et al.
Plant physiology, 151(3), 1646-1657 (2009-09-17)
In vivo (31)P-NMR analyses showed that the phosphate (Pi) concentration in the cytosol of sycamore (Acer pseudoplatanus) and Arabidopsis (Arabidopsis thaliana) cells was much lower than the cytoplasmic Pi concentrations usually considered (60-80 mum instead of >1 mm) and that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service