Skip to Content
Merck
All Photos(3)

Key Documents

287059

Sigma-Aldrich

L-Prolinamide

98%, for peptide synthesis

Synonym(s):

(2S)-2-Carbamoylpyrrolidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10N2O
CAS Number:
Molecular Weight:
114.15
Beilstein:
80807
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

product name

L-Prolinamide, 98%

Assay

98%

optical activity

[α]20/D −106°, c = 1 in ethanol

reaction suitability

reaction type: solution phase peptide synthesis

mp

95-97 °C (lit.)

application(s)

peptide synthesis

SMILES string

NC(=O)[C@@H]1CCCN1

InChI

1S/C5H10N2O/c6-5(8)4-2-1-3-7-4/h4,7H,1-3H2,(H2,6,8)/t4-/m0/s1

InChI key

VLJNHYLEOZPXFW-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Denis R St Laurent et al.
Bioorganic & medicinal chemistry letters, 22(19), 6063-6066 (2012-09-11)
In a previous disclosure,(1) we reported the dimerization of an iminothiazolidinone to form 1, a contributor to the observed inhibition of HCV genotype 1b replicon activity. The dimer was isolated via bioassay-guided fractionation experiments and shown to be a potent
Tor E Kristensen et al.
Organic letters, 11(14), 2968-2971 (2009-06-16)
A completely non-chromatographic and highly large-scale adaptable synthesis of acrylic polymer beads containing proline and prolineamides has been developed. Novel monomeric proline (meth)acrylates are prepared from hydroxyproline in only one step. Free-radical copolymerization then gives solid-supported proline organocatalysts directly in
Highly enantioselective strecker reaction of ketoimines catalyzed by an organocatalyst from (S)-BINOL and L-prolinamide.
Zongrui Hou et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 14(15), 4484-4486 (2008-04-11)
Hidenobu Komeda et al.
European journal of biochemistry, 271(8), 1465-1475 (2004-04-07)
An amidase acting on (R,S)-piperazine-2-tert-butylcarboxamide was purified from Pseudomonas azotoformans IAM 1603 and characterized. The enzyme acted S-stereoselectively on (R,S)-piperazine-2-tert-butylcarboxamide to yield (S)-piperazine-2-carboxylic acid. N-terminal and internal amino acid sequences of the enzyme were determined. The gene encoding the S-stereoselective
Carlos R Baiz et al.
Chemical reviews, 120(15), 7152-7218 (2020-07-01)
Vibrational spectroscopy is an essential tool in chemical analyses, biological assays, and studies of functional materials. Over the past decade, various coherent nonlinear vibrational spectroscopic techniques have been developed and enabled researchers to study time-correlations of the fluctuating frequencies that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service