Skip to Content
Merck
All Photos(1)

Documents

282855

Sigma-Aldrich

3-Methyl-2,4-pentanedione, mixture of tautomers

technical grade, 85%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH(CH3)COCH3
CAS Number:
Molecular Weight:
114.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

85%

form

liquid

refractive index

n20/D 1.442 (lit.)

bp

172-174 °C (lit.)

density

0.981 g/mL at 25 °C (lit.)

SMILES string

CC(C(C)=O)C(C)=O

InChI

1S/C6H10O2/c1-4(5(2)7)6(3)8/h4H,1-3H3

InChI key

GSOHKPVFCOWKPU-UHFFFAOYSA-N

General description

Peroxynitrite promoted aerobic oxidation of 3-methyl-2,4-pentanedione has been reported. The kinetics of the reaction of OH radicals with 3-methyl-2,4-pentanedione has been investigated in the gas-phase using a pulsed laser photolysis-laser induced fluorescence technique. 3-methyl-2,4-pentanedione participates in asymmetric substitution of 1,3-diphenyl-2-propenyl acetate catalyzed by amphiphilic resin-supported monodentate phosphine ligands.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F S Knudsen et al.
Chemical research in toxicology, 13(5), 317-326 (2000-05-17)
Peroxynitrite is shown here to promote the aerobic oxidation of isobutanal (IBAL) and 3-methyl-2,4-pentanedione (MP) in a pH 7.2 phosphate buffer into acetone plus formate and biacetyl plus acetate, respectively. These products are expected from dioxetane intermediates, whose thermolysis is
Rate coefficients for the reactions of OH radicals with the keto/enol tautomers of 2, 4-pentanedione and 3-methyl-2, 4-pentanedione, allyl alcohol and methyl vinyl ketone using the enols and methyl nitrite as photolytic sources of OH.
Holloway A-L, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 176(1), 183-190 (2005)
Palladium-catalyzed asymmetric allylic substitution in aqueous media using amphiphilic resin-supported MOP ligands.
Uozumi Y, et al.
Tetrahedron Letters, 39(45), 8303-8306 (1998)
Marzena Fandzloch et al.
Journal of inorganic biochemistry, 176, 144-155 (2017-09-15)
Tropical diseases currently constitute a major health problem and thus a challenge in the field of drug discovery. The current treatments show serious disadvantages due to cost, toxicity, long therapy duration and resistance, and the use of metal complexes as
Jiyuan Jin et al.
Chemosphere, 223, 628-635 (2019-02-25)
Quinones are components of electron transport chains in photosynthesis and respiration. Acetylacetone (AA), structurally similar to benzoquinone (BQ) for the presence of two identical carbonyl groups, has been reported as a quinone-like electron shuttle. Both BQ and AA are important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service