Skip to Content
Merck
All Photos(1)

Documents

281697

Sigma-Aldrich

(R)-(−)-2-Pyrrolidinemethanol

99%

Synonym(s):

(R)-(−)-2-(Hydroxymethyl)pyrrolidine, D-Prolinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO
CAS Number:
Molecular Weight:
101.15
Beilstein:
1523669
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

optical activity

[α]20/D −31°, c = 1 in toluene

refractive index

n20/D 1.4849 (lit.)

bp

74-76 °C/2 mmHg (lit.)

density

1.025 g/mL at 25 °C (lit.)

SMILES string

OC[C@H]1CCCN1

InChI

1S/C5H11NO/c7-4-5-2-1-3-6-5/h5-7H,1-4H2/t5-/m1/s1

InChI key

HVVNJUAVDAZWCB-RXMQYKEDSA-N

Application

Versatile reagent for asymmetric synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Dei et al.
Bioorganic & medicinal chemistry, 11(14), 3153-3164 (2003-06-24)
Four out of the eight possible stereoisomers of 1-methyl-2-(2-methyl-1,3-dioxolan-4-yl)pyrrolidine, 1-methyl-2-(2-methyl-1,3-oxathiolan-5-yl)pyrrolidine and the corresponding iodomethylates have been synthesised. They were formally derived from hybridisation of potent though unselective agonists studied before, such as 1,3-dioxolane 1 and 1,3-oxathiolane 2, with the structure
Tetrahedron Asymmetry, 15, 519-523 (2004)
Emily M Christensen et al.
The Journal of biological chemistry, 295(52), 18316-18327 (2020-10-29)
Pyrroline-5-carboxylate reductase 1 (PYCR1) catalyzes the biosynthetic half-reaction of the proline cycle by reducing Δ1-pyrroline-5-carboxylate (P5C) to proline through the oxidation of NAD(P)H. Many cancers alter their proline metabolism by up-regulating the proline cycle and proline biosynthesis, and knockdowns of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service