Skip to Content
Merck
All Photos(1)

Key Documents

275735

Sigma-Aldrich

2,5-Di-tert-butylaniline

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H3NH2
CAS Number:
Molecular Weight:
205.34
Beilstein:
2691916
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
powder
Assay:
99%

Quality Level

Assay

99%

form

powder

mp

103-106 °C (lit.)

SMILES string

CC(C)(C)c1ccc(c(N)c1)C(C)(C)C

InChI

1S/C14H23N/c1-13(2,3)10-7-8-11(12(15)9-10)14(4,5)6/h7-9H,15H2,1-6H3

InChI key

NUZVLYNISQOZOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,5-Di-tert-butylaniline has been used in the preparation of:
  • new rigid bidentate nitrogen ligands 1,2-bis[(2,5-di-tert-butylphenyl)imino]acenaphthene (dtb-BIAN)
  • N-(2,5-di-tert-butylphenyl)perylene-3,4-dicarboximide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New perylenes for dye sensitization of TiO2.
Ferrere S and Gregg BA.
New. J. Chem., 26(9), 1155-1160 (2002)
Monomeric Magnesium and Calcium Complexes containing the Rigid, Dianionic 1, 2-Bis [(2, 5-di-tert-butylphenyl) imino] acenaphthene (dtb-BIAN) and 1, 2-Bis [(2-biphenyl) imino] acenaphthene (bph-BIAN) Ligands.
Fedushkin IL, et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 630(4), 501-507 (2004)
Christopher Elam et al.
European journal of medicinal chemistry, 46(5), 1512-1523 (2011-03-01)
Two screening protocols based on recursive partitioning and computational ligand docking methodologies, respectively, were employed for virtual screens of a compound library with 345,000 entries for novel inhibitors of the enzyme sarco/endoplasmic reticulum calcium ATPase (SERCA), a potential target for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service