Skip to Content
Merck
All Photos(1)

Key Documents

274879

Sigma-Aldrich

1,4-Cyclohexanedione monoethylene acetal

97%

Synonym(s):

1,4-Dioxaspiro[4.5]decan-8-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O3
CAS Number:
Molecular Weight:
156.18
Beilstein:
117631
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

70-73 °C (lit.)

functional group

ether
ketal
ketone

SMILES string

O=C1CCC2(CC1)OCCO2

InChI

1S/C8H12O3/c9-7-1-3-8(4-2-7)10-5-6-11-8/h1-6H2

InChI key

VKRKCBWIVLSRBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in a study of microwave-assisted reductive amination.
Via a novel aromatization process, this bicyclic frame can be converted into a series of 5-alkoxytryptamine derivatives including serotonin, melatonin and bufotenin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synlett, 2444-2444 (2006)
Synthetic Communications, 24, 799-799 (1994)
Tetrahedron, 50, 731-731 (1994)
Tetrahedron Letters, 45, 4859-4859 (2004)
Aldrichimica Acta, 12, 40-40 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service