Skip to Content
Merck
All Photos(1)

Key Documents

274100

Sigma-Aldrich

cis,cis-1,3-Cyclooctadiene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12
CAS Number:
Molecular Weight:
108.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

liquid

refractive index

n20/D 1.493 (lit.)

bp

55 °C/34 mmHg (lit.)

mp

−53-−51 °C (lit.)

density

0.869 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1CCC=CC=CC1

InChI

1S/C8H12/c1-2-4-6-8-7-5-3-1/h1-4H,5-8H2/b3-1-,4-2-

InChI key

RRKODOZNUZCUBN-CCAGOZQPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The photosensitized cyclization of cis,cis-1,3-cyclooctadiene to bicyclo[4.2.0]oct-7-ene was studied. It proceeded by first isomerization of the diene to its cis, trans isomer followed by cyclization[1].

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photosensitized isomerization of 1, 3-cyclooctadienes and conversion to bicyclo [4.2. 0] oct-7-ene.
Journal of the American Chemical Society, 89(1), 112-114 (1967)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service