Skip to Content
Merck
All Photos(1)

Key Documents

272973

Sigma-Aldrich

1,1-Dichloropropane

97%

Synonym(s):

Propylidene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CHCl2
CAS Number:
Molecular Weight:
112.99
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.428 (lit.)

bp

88 °C (lit.)

solubility

organic solvents: soluble(lit.)
water: very slightly soluble(lit.)

density

1.13 g/mL at 25 °C (lit.)

SMILES string

CCC(Cl)Cl

InChI

1S/C3H6Cl2/c1-2-3(4)5/h3H,2H2,1H3

InChI key

WIHMGGWNMISDNJ-UHFFFAOYSA-N

Application

1,1-Dichloropropane was used in the synthesis of chloropropenes by undergoing flash vacuum pyrolysis over magnesium.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F - closed cup

Flash Point(C)

7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flash vacuum pyrolysis over magnesium. Part 1. Pyrolysis of benzylic, other aryl/alkyl and aliphatic halides.
Journal of the Chemical Society. Perkin Transactions 1, 3, 402-415 (2002)
Na-Hee Lee et al.
Cell death & disease, 9(7), 724-724 (2018-06-22)
The toxicological evaluation of potential drug candidates is very important in the preclinical phase of drug development. Toxic materials may cause serious decline in stem cell function and loss of stemness. Indeed, we found that toxic exposure more profoundly suppressed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service