Skip to Content
Merck
All Photos(2)

Key Documents

270784

Sigma-Aldrich

4-(Chloromethyl)benzoyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2C6H4COCl
CAS Number:
Molecular Weight:
189.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

126-128 °C/6 mmHg (lit.)

mp

30-32 °C (lit.)

functional group

acyl chloride
chloro

SMILES string

ClCc1ccc(cc1)C(Cl)=O

InChI

1S/C8H6Cl2O/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4H,5H2

InChI key

RCOVTJVRTZGSBP-UHFFFAOYSA-N

Application

4-(Chloromethyl)benzoyl chloride was used as an atom transfer radical polymerization (ATRP) initiator in the growth of polyacrylamide (PAAm) brushes from silicon wafers. It was also used as a building block in a microwave-assisted, solid-phase synthesis of imatinib, an anticancer agent.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Song Yi Lee et al.
ACS applied materials & interfaces, 10(43), 36628-36640 (2018-10-10)
CD44 receptor targeting and lipid rafts destroying nanoassembly (NA) was developed for breast cancer therapy. Methyl-β-cyclodextrin (MbCD), as a cholesterol depletion moiety, was conjugated to hyaluronic acid-ceramide (HACE) structure via an ester linkage. HACE-MbCD NA with 198 nm hydrodynamic size
Tetrahedron Letters, 48, 3455-3455 (2007)
Irina Cringus-Fundeanu et al.
Langmuir : the ACS journal of surfaces and colloids, 23(9), 5120-5126 (2007-03-29)
A method is presented to prevent microbial adhesion to solid surfaces exploiting the unique properties of polymer brushes. Polyacrylamide (PAAm) brushes were grown from silicon wafers by atom transfer radical polymerization (ATRP) using a three-step reaction procedure consisting of immobilization
Andrej Babič et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 24(6), 1348-1357 (2017-11-10)
Gadolinium-loaded nanomicelles show promise as future magnetic resonance imaging (MRI) contrast agents (CAs). Their increased size and high gadolinium (Gd) loading gives them an edge in proton relaxivity over smaller molecular Gd-complexes. Their size and stealth properties are fundamental for
Ali Bilgic et al.
Journal of fluorescence, 30(4), 867-881 (2020-06-05)
In this study, we developed two different very sensitive magnetite fluorescent Fe3O4@SiO2-TPED-BODIPY and Fe3O4@SiO2-TMPTA-BODIPY nano-sensors for the selective detection of Cr(VI) ions. The Cr(VI) metal ions sensing is based on the fluorescent quenching of BODIPY functionalized with Fe3O4@SiO2-TPED and Fe3O4@SiO2-TMPTA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service