Skip to Content
Merck
All Photos(1)

Key Documents

269190

Sigma-Aldrich

Methanesulfonic anhydride

97%

Synonym(s):

Mesyl anhydride, Mesylic anhydride, Methanesulfonyl anhydride, Methylsulfonyl methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3SO2)2O
CAS Number:
Molecular Weight:
174.20
Beilstein:
972316
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.3 (vs air)

Quality Level

Assay

97%

form

solid

bp

125 °C/4 mmHg (lit.)

mp

64-67 °C (lit.)

functional group

sulfonic acid

SMILES string

CS(=O)(=O)OS(C)(=O)=O

InChI

1S/C2H6O5S2/c1-8(3,4)7-9(2,5)6/h1-2H3

InChI key

IZDROVVXIHRYMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methanesulfonic anhydride is an organic compound that is commonly used as a reagent for the preparation of methyl sulfonates (mesylates). It is also used in the synthesis of methanesulfonamides using various amine precursors. Additionally, the mixture of methanesulfonic anhydride and dimethyl sulfoxide can be used as an oxidizing agent for the oxidation of primary and secondary alcohols to aldehydes and ketones.

Application

Methanesulfonic anhydride can be used as an amine base to synthesize glycosyl mesylates in situ from the corresponding free hemiacetals under mild conditions.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark C Wilkinson
Organic letters, 13(9), 2232-2235 (2011-03-29)
The utility of methanesulfonic anhydride for promoting the Friedel-Crafts acylation reaction of aryl and alkyl carboxylic acids is disclosed. This reagent allows the preparation of aryl ketones in good yield with minimal waste containing no metallic or halogenated components, clearly
Keith Smith et al.
Organic & biomolecular chemistry, 2(21), 3150-3154 (2004-10-27)
Regioselective methanesulfonylation of simple aromatics using methanesulfonic anhydride can be achieved over zeolite catalysts. For example, methanesulfonylation of toluene over various cation-exchanged zeolite beta catalysts affords higher para-selectivity in the synthesis of methyl tolyl sulfone than standard Friedel-Crafts methanesulfonylation utilising
Mohammad Hasani et al.
The journal of physical chemistry. B, 123(8), 1815-1821 (2019-02-20)
Protic ionic liquids (PILs) are made by proton transfer from a Brønsted acid to a base and are of interest for their solvent and electrolyte properties such as high ionic conductivity. Unfortunately, many PILs have been misnamed, because their ionic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service