Skip to Content
Merck
All Photos(1)

Key Documents

265128

Sigma-Aldrich

5-Fluoroindole-2-carboxylic acid

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6FNO2
CAS Number:
Molecular Weight:
179.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

259 °C (dec.) (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)c1cc2cc(F)ccc2[nH]1

InChI

1S/C9H6FNO2/c10-6-1-2-7-5(3-6)4-8(11-7)9(12)13/h1-4,11H,(H,12,13)

InChI key

WTXBRZCVLDTWLP-UHFFFAOYSA-N

General description

5-Fluoroindole-2-carboxylic acid is an antagonist of the glycine site within the NMDA (N-methyl-D-aspartate) receptor complex.

Application

Reactant for the synthesis of:
  • Fungicidal agents
  • Antitumor agents
  • 2,3-dioxygenase (IDO) inhibitors
  • Factor Xa inhibitors
  • Enantioselective D3 receptor antagonists
  • Ligands for hFPRL1 (or ALXR) receptor in inflammation
  • Antibacterial agents
  • Inhibitors of hepatitis C virus NS3·4A protease

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Kapoor et al.
Clinical and experimental pharmacology & physiology, 25(3-4), 216-219 (1998-05-20)
1. The effects of the specific N-methyl-D-aspartate (NMDA)-glycine site antagonist 5-fluoro indole-2-carboxylic acid (FICA) and NMDA, microinjected into the vasodepressor caudal ventrolateral medulla, were compared in spontaneously hypertensive rats (SHR) and in Wistar-Kyoto (WKY) rats. 2. 5-Fluoro indole-2-carboxylic acid elicited
T J Grudt et al.
Molecular pharmacology, 37(4), 477-481 (1990-04-01)
Whole-cell and single-channel patch-clamp recordings from hippocampal neurons in culture have been used to study the receptor channel selectivity of the glutamate analog quisqualate. The dose-response relationship of quisqualate acting at the N-methyl-D-aspartate (NMDA) receptor was measured as that portion
R Kamiński et al.
Journal of neural transmission (Vienna, Austria : 1996), 105(2-3), 133-146 (1998-07-11)
5-Fluoroindole-2-carboxylic acid, an antagonist of the glycine site within the NMDA receptor complex, administered intraperitoneally in doses of 150 and 200 mg/kg, 120 min before electroconvulsions, significantly raised the convulsive threshold from 6.8 to 7.9 and 8.3 mA, respectively. At
R Bakshi et al.
Neuroscience letters, 110(1-2), 113-117 (1990-03-02)
Intrathecal (i.t.) administration of the opioid dynorphin causes neurological dysfunction and tissue damage. It has been suggested that these effects of dynorphin may be mediated, in part, by N-methyl-D-aspartate (NMDA) receptors. In the present studies, recently developed compounds that block
Chung-Yi Wu et al.
Chemistry & biology, 13(3), 261-268 (2006-04-28)
Severe acute respiratory syndrome (SARS) is caused by a newly emerged coronavirus that infected more than 8000 individuals and resulted in more than 800 fatalities in 2003. Currently, there is no effective treatment for this epidemic. SARS-3CL(pro) has been shown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service