Skip to Content
Merck
All Photos(3)

Key Documents

263567

Sigma-Aldrich

1,4-Bis(trimethylsilyl)butadiyne

98%, stable crystalline form of butadiyne

Synonym(s):

1,4-Bis(trimethylsilyl)-1,3-butadiyne, BTMSBD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiC≡CC≡CSi(CH3)3
CAS Number:
Molecular Weight:
194.42
Beilstein:
1758268
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

refractive index

n20/D 1.384 (lit.)

mp

111-113 °C (lit.)

density

0.974 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[Si](C)(C)C#CC#C[Si](C)(C)C

InChI

1S/C10H18Si2/c1-11(2,3)9-7-8-10-12(4,5)6/h1-6H3

InChI key

LBNVCJHJRYJVPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,4-Bis(trimethylsilyl)butadiyne can be used as a reagent to prepare:
  • 1,1,3,4-Tetrasilyl-substituted 1,3-butadienes or 1,1,3,4-tetrasilyl-substituted 1,2-butadienes by hydrosilylation reaction using various hydridosilanes and catalysts.
  • Glycosylated oligo(ethynylene)s using the Negishi reaction.
  • (±) Falcarinol, a polyacetylene class of fatty alcohol.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A short synthesis of (+) and (-)-falcarinol
McLaughlin NP, et al.
Tetrahedron, 66, 9681-9687 (2010)
Manuel M Bentlohner et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(67), 17089-17094 (2017-09-15)
We report on the synthesis and structure, as well as on the mechanism of formation of the [Ge
Hydrosilylation of 1, 4-Bis (trimethylsilyl) butadiyne and Silyl-Substituted Butenynes.
Kusumoto T, et al.
Bulletin of the Chemical Society of Japan, 65(5), 1280-1290 (1992)
Manuel M Bentlohner et al.
Angewandte Chemie (International ed. in English), 54(12), 3748-3753 (2015-02-04)
The accessibility of triads with deltahedral Zintl clusters in analogy to fullerene-linker-fullerene triads is another example for the close relationship between fullerenes and Zintl clusters. The compound {[K(2.2.2-crypt)]4[RGe9-CH=CH-CH=CH-Ge9R]}(toluene)2 (R=(2Z,4E)-7-amino-5-aza-hepta-2,4-dien-2-yl), containing two deltahedral [Ge9] clusters linked by a conjugated (1Z,3Z)-buta-1,3-dien-1,4-diyl bridge
Tobias N Hoheisel et al.
Organic letters, 10(20), 4525-4528 (2008-09-25)
A convenient and efficient sp-sp carbon heterocoupling protocol based on the Negishi reaction was developed, in which the required zinc diacetylide was generated from 1,4-bis(trimethylsilyl)butadiyne in situ and reacted with a bromoacetylene in apolar solvent mixtures. The method has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service