Skip to Content
Merck
All Photos(3)

Documents

263265

Sigma-Aldrich

Gallium

99.99% trace metals basis

Synonym(s):

Elemental gallium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ga
CAS Number:
Molecular Weight:
69.72
MDL number:
UNSPSC Code:
12141715
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

(metallic liquid or solid (ambient temp dependent))

resistivity

25.795 μΩ-cm, 30°C

bp

2403 °C (lit.)

mp

29.8 °C (lit.)

density

5.904 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[Ga]

InChI

1S/Ga

InChI key

GYHNNYVSQQEPJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gallium is a rareearth element that is widely used in smart electronic devices and integratedcircuits as it provides the advantages of low energy consumption and highcomputational speeds. Due to its high boiling point and low melting point, itexhibits a wide temperature range in the liquid state. It tends to formsulfate, chloride, and phosphate complexes due to its high charge. Thecompounds of gallium find applications in advanced semiconductors and LEDchips.

Application

Gallium can be used as a precursor to synthesize:
  • Gallium nitride and other Ga-based compounds for optoelectronic applications.
  • Sulfur/gallium hybrid cathode material for lithium-sulfur batteries. The surface of Ga metal promotes the redox reaction of lithium polysulfides due to its electrocatalytic effect.
  • Ga-based liquid alloys for the fabrication offlexible electronic devices.

Features and Benefits

  • Excellent conductor of heat and electricity
  • Wide temperature range in the liquid state
  • Lowvapor pressure at high temperature

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth M Carnicom et al.
Dalton transactions (Cambridge, England : 2003), 46(41), 14158-14163 (2017-10-07)
Previously unreported σ-phases in the ternary niobium-based systems Nb-X-Ga for X = Ru, Rh, Pd, Ir, Pt, Au, and Nb-X-Al for X = Ir, and Pt are presented, prepared by arc-melting followed by annealing at 1000 °C for 1 week.
Sayan Sarkar et al.
Scientific reports, 9(1), 8180-8180 (2019-06-05)
Recent advances in high performance thermoelectric materials have garnered unprecedented attention owing to their capability of direct transformation of heat energy to useful electricity. Copper Telluride (Cu2Te), a member of the chalcogenide family has emerged as a state-of-the-art thermoelectric material
Prasana Sahoo et al.
Biosensors & bioelectronics, 44, 164-170 (2013-02-22)
We demonstrate a very simple and generic protocol for ultrasensitive in-situ label-free detection of DNA hybridization using third generation poly(amidoamine)dendrimer (G3-PAMAM) functionalized GaN nanowires (NWs). PAMAM modified GaN NWs provides large density of docking site to immobilize significant number of
Michael A Jakupec et al.
Metal ions in biological systems, 42, 425-462 (2004-06-23)
Gallium has been the second metal to show activity against malignant tumors in humans soon after the establishment of platinum drugs in routine clinical practice. It has the unique property of inhibiting tumor growth as a simple cation, mainly because
Michael A Jakupec et al.
Current topics in medicinal chemistry, 4(15), 1575-1583 (2004-12-08)
The trivalent gallium cation is capable of inhibiting tumor growth, mainly because of its resemblance to ferric iron. It affects cellular acquisition of iron by binding to transferrin, and it interacts with the iron-dependent enzyme ribonucleotide reductase, resulting in reduced

Articles

Solid state and materials chemistry have made a tremendous impact and have experienced growth in recent years, particularly for rare earthcontaining materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service