Skip to Content
Merck
All Photos(2)

Key Documents

254142

Sigma-Aldrich

Bismuth(III) chloride

99.99% trace metals basis

Synonym(s):

Bismuth trichloride, Trichlorobismuth

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BiCl3
CAS Number:
Molecular Weight:
315.34
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
solid
Pricing and availability is not currently available.

grade

for analytical purposes

Assay

99.99% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: bismuth

impurities

≤150.0 ppm Trace Metal Analysis

bp

447 °C (lit.)

mp

230-232 °C (lit.)

SMILES string

Cl[Bi](Cl)Cl

InChI

1S/Bi.3ClH/h;3*1H/q+3;;;/p-3

InChI key

JHXKRIRFYBPWGE-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Bismuth(III) chloride is a Lewis acid catalyst and a common source of Bi3+ ions used in various organic transformations and synthesis of bismuth nanomaterials.[1] [2]

Application

Bismuth(III) chloride can be used:
  • As a precursor to prepare polyaniline and bismuth(III) telluride nanohybrids with high thermoelectric performance.[1]
  • As a starting material to synthesize visible light absorbing double perovskite semiconductors, such as Cs2AgBiBr6 and Cs2AgBiCl6, eco-friendly alternative to the lead halide perovskites.[3]
  • To form a protective layer on anode materials.[4]
It can also be used as an efficient catalyst for green synthesis of:
  • Chalcones by Claisen–Schmidt reaction under solvent-free conditions.[5]
  • One-pot three-component synthesis of Propargylamines.[2]
  • Coumarins via ultrasound‐assisted Pechmann condensation of phenols with β‐ketoesters.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A bismuth-based protective layer for magnesium metal anode in noncorrosive electrolytes
Yimin Zhao, et al.
ACS Energy Letters, 6, 2594-2601 (2021)
Organic-inorganic nanohybrids as novel thermoelectric materials: hybrids of polyaniline and bismuth (III) telluride nanoparticles
Naoki Toshima, et al.
Journal of Electronic Materials, 40, 898-902 (2011)
A simple and efficient one-pot three-component synthesis of propargylamines using bismuth (III) chloride
Abbas Teimouri, et al.
Bulletin of the Korean Chemical Society,, 33, 1556-1560 (2012)
An efficient green protocol for the synthesis of chalcones by a Claisen-Schmidt reaction using bismuth (III) chloride as a catalyst under solvent-free condition
Dhruva Kumar, et al.
Green Chemistry Letters and Reviews, 3, 283-286 (2010)
Cs2AgBiX6 (X= Br, Cl): new visible light absorbing, lead-free halide perovskite semiconductors
Eric T McClure, et al.
Chemistry of Materials, 28, 1348-1354 (2016)

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service