Skip to Content
Merck
All Photos(1)

Key Documents

249645

Sigma-Aldrich

2-Acetyl-3-methylthiophene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8OS
CAS Number:
Molecular Weight:
140.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.562 (lit.)

bp

98-99 °C/14 mmHg (lit.)

density

1.124 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1sccc1C

InChI

1S/C7H8OS/c1-5-3-4-9-7(5)6(2)8/h3-4H,1-2H3

InChI key

YBJDKNXEWQSGEL-UHFFFAOYSA-N

Application

2-Acetyl-3-methylthiophene has been used in:
  • automated parallel synthesis of combinatorial arrays of individual chalcone derivatives
  • synthesis of 2-hydroxy-4-(3-methylthiophen-2-yl)-4-oxobut-2-enoic acid

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aaditya Bhatt et al.
European journal of medicinal chemistry, 46(10), 5138-5145 (2011-09-07)
Hepatitis C virus (HCV) NS5B polymerase is a key target for anti-HCV therapeutics development. Here we report the synthesis and biological evaluation of a new series of α,γ-diketo acids (DKAs) as NS5B polymerase inhibitors. We initiated structure-activity relationship (SAR) optimization
Automated parallel synthesis of chalcone-based screening libraries.
Powers DG, et al.
Tetrahedron, 54(16), 4085-4096 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service