Skip to Content
Merck
All Photos(1)

Key Documents

249084

Sigma-Aldrich

cis-11-Hexadecenal

95%

Synonym(s):

(Z)-11-HDAL

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH=CH(CH2)9CHO
CAS Number:
Molecular Weight:
238.41
Beilstein:
1867354
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
95%

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.455 (lit.)

density

0.963 g/mL at 20 °C (lit.)

functional group

aldehyde

storage temp.

2-8°C

SMILES string

[H]C(=O)CCCCCCCCC\C=C/CCCC

InChI

1S/C16H30O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h5-6,16H,2-4,7-15H2,1H3/b6-5-

InChI key

AMTITFMUKRZZEE-WAYWQWQTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

cis-11-Hexadecenal is the major pheromone component detected in the whole-animal male Helicoverpa zea antennal preparations.

Application

cis-11-Hexadecenal has been used in the preparation of fatty aldehyde semicarbazone derivatives during their analysis by liquid chromatography-electrospray ionization-tandem mass spectrometery.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew J Myrick et al.
Biosensors & bioelectronics, 31(1), 197-204 (2011-11-25)
A new method that can improve gas-chromatography-electroantennographic detection (GC-EAD) by orders of magnitude through a technique known as chopper stabilization combined with matched filtering in colored noise is presented. The EAD is a physiological recording from the antenna of an
Evgeny V Berdyshev et al.
Analytical biochemistry, 408(1), 12-18 (2010-09-02)
Sphingosine-1-phosphate (S1P) is a sphingolipid signaling molecule crucial for cell survival and proliferation. S1P-mediated signaling is largely controlled through its biosynthesis and degradation, and S1P lyase (S1PL) is the only known enzyme that irreversibly degrades sphingoid base-1-phosphates to phosphoethanolamine and
Hong-Lei Wang et al.
Insect biochemistry and molecular biology, 35(6), 575-583 (2005-04-29)
Two Helicoverpa species, H. armigera and H. assulta use (Z)-11-hexadecenal and (Z)-9-hexadecenal as their sex attractant pheromone components but in opposite ratios. Since both female and male interspecific hybrids produced by female H. assulta and male H. armigera have been
E E Blatter et al.
The Biochemical journal, 272(2), 351-358 (1990-12-01)
A major component of the sex pheromone from the tobacco budworm moth Heliothis virescens is a C16 straight-chain aldehyde with a single unsaturation at the eleventh position. The sex pheromones are inactivated when metabolized to their corresponding acids by insect
Carmen Quero et al.
Journal of agricultural and food chemistry, 51(10), 2987-2991 (2003-05-02)
The sex pheromone of the oak processionary moth Thaumetopoea processionea has been characterized from female gland extracts as a mixture of (Z,Z)-11,13-hexadecadienyl acetate (1), (E,Z)-11,13-hexadecadienyl acetate (3) and (Z,Z)-11,13-hexadecadienol (2) in 88:7:5 ratio. The amount of the major compound 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service